  • Search Results
  • Amino Acid Transporter Slc38a5 Controls Glucagon Receptor Inhibition-Induced Pancreatic α Cell Hyperplasia in Mice.
Amino Acid Transporter Slc38a5 Controls Glucagon Receptor Inhibition-Induced Pancreatic α Cell Hyperplasia in Mice.
Cell metabolism (2017-06-08)
Jinrang Kim, Haruka Okamoto, ZhiJiang Huang, Guillermo Anguiano, Shiuhwei Chen, Qing Liu, Katie Cavino, Yurong Xin, Erqian Na, Rachid Hamid, Joseph Lee, Brian Zambrowicz, Roger Unger, Andrew J Murphy, Yan Xu, George D Yancopoulos, Wen-Hong Li, Jesper Gromada
ABSTRACT
Glucagon supports glucose homeostasis by stimulating hepatic gluconeogenesis, in part by promoting the uptake and conversion of amino acids into gluconeogenic precursors. Genetic disruption or pharmacologic inhibition of glucagon signaling results in elevated plasma amino acids and compensatory glucagon hypersecretion involving expansion of pancreatic α cell mass. Recent findings indicate that hyperaminoacidemia triggers pancreatic α cell proliferation via an mTOR-dependent pathway. We confirm and extend these findings by demonstrating that glucagon pathway blockade selectively increases expression of the sodium-coupled neutral amino acid transporter Slc38a5 in a subset of highly proliferative α cells and that Slc38a5 controls the pancreatic response to glucagon pathway blockade; most notably, mice deficient in Slc38a5 exhibit markedly decreased α cell hyperplasia to glucagon pathway blockade-induced hyperaminoacidemia. These results show that Slc38a5 is a key component of the feedback circuit between glucagon receptor signaling in the liver and amino-acid-dependent regulation of pancreatic α cell mass in mice.
Sigma-Aldrich
色氨酸 -L, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
柠檬酸盐缓冲液,pH 6.0,10×,抗原修复液
Sigma-Aldrich
L-脯氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
L-异亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-天冬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
单克隆抗胰高血糖素 小鼠抗, clone K79bB10, ascites fluid
Sigma-Aldrich
L-氨基酸定量检测试剂盒, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
抗生长抑素 兔抗, affinity isolated antibody
Sigma-Aldrich
Anti-Pancreatic polypeptide/PPY antibody produced in goat, affinity isolated antibody, buffered aqueous solution