  • Search Results
  • Defining the Architecture of the Core Machinery for the Assembly of Fe-S Clusters in Human Mitochondria
Defining the Architecture of the Core Machinery for the Assembly of Fe-S Clusters in Human Mitochondria
Methods in Enzymology (2017)