  • Search Results
  • A Dietary Fiber-Deprived Gut Microbiota Degrades the Colonic Mucus Barrier and Enhances Pathogen Susceptibility.
A Dietary Fiber-Deprived Gut Microbiota Degrades the Colonic Mucus Barrier and Enhances Pathogen Susceptibility.
Cell (2016-11-20)
Mahesh S Desai, Anna M Seekatz, Nicole M Koropatkin, Nobuhiko Kamada, Christina A Hickey, Mathis Wolter, Nicholas A Pudlo, Sho Kitamoto, Nicolas Terrapon, Arnaud Muller, Vincent B Young, Bernard Henrissat, Paul Wilmes, Thaddeus S Stappenbeck, Gabriel Núñez, Eric C Martens
ABSTRACT
Despite the accepted health benefits of consuming dietary fiber, little is known about the mechanisms by which fiber deprivation impacts the gut microbiota and alters disease risk. Using a gnotobiotic mouse model, in which animals were colonized with a synthetic human gut microbiota composed of fully sequenced commensal bacteria, we elucidated the functional interactions between dietary fiber, the gut microbiota, and the colonic mucus barrier, which serves as a primary defense against enteric pathogens. We show that during chronic or intermittent dietary fiber deficiency, the gut microbiota resorts to host-secreted mucus glycoproteins as a nutrient source, leading to erosion of the colonic mucus barrier. Dietary fiber deprivation, together with a fiber-deprived, mucus-eroding microbiota, promotes greater epithelial access and lethal colitis by the mucosal pathogen, Citrobacter rodentium. Our work reveals intricate pathways linking diet, the gut microbiome, and intestinal barrier dysfunction, which could be exploited to improve health using dietary therapeutics.
Sigma-Aldrich
盐酸, ACS reagent, 37%
Sigma-Aldrich
十二烷基硫酸钠, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
生物素, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
硫酸镁, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
D- (-)-果糖, ≥99%
Sigma-Aldrich
氢氧化钠, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
乙酸钠, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-脯氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
维生素 B 12, ≥98%
Sigma-Aldrich
硼酸, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
L -赖氨酸, ≥98% (TLC)
Sigma-Aldrich
L-异亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
海藻酸 钠盐, powder
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
叶酸, ≥97%
Sigma-Aldrich
L-苏氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
烟酸, ≥98%
Sigma-Aldrich
L -缬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
D -(+)-半乳糖, ≥98%
Sigma-Aldrich
(−)-核黄素, meets USP testing specifications
Sigma-Aldrich
D-(+)-木糖, BioXtra, ≥99%
Sigma-Aldrich
4-硝基苯 α-L-岩藻糖苷, ≥98% (TLC)