  • Search Results
  • Method for Multiplexing CRISPR/Cas9 in Saccharomyces cerevisiae Using Artificial Target DNA Sequences.
Method for Multiplexing CRISPR/Cas9 in Saccharomyces cerevisiae Using Artificial Target DNA Sequences.
Bio-protocol (2017-10-31)
Rachael M Giersch, Gregory C Finnigan
ABSTRACT
Genome manipulation has become more accessible given the advent of the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) editing technology. The Cas9 endonuclease binds a single stranded (single guide) RNA (sgRNA) fragment that recruits the complex to a corresponding genomic target sequence where it induces a double stranded break. Eukaryotic repair systems allow for the introduction of exogenous DNA, repair of existing mutations, or deletion of endogenous gene products. Targeting of Cas9 to multiple genomic positions (termed 'multiplexing') is achieved by the expression of multiple sgRNAs within the same nucleus. However, an ongoing concern of the CRISPR field has been the accidental targeting of Cas9 to alternative ('off-target') DNA locations within a genome. We describe the use (dubbed Multiplexing of Cas9 at Artificial Loci) of installed artificial Cas9 target sequences into the yeast genome that allow for (i) multiplexing with a single sgRNA; (ii) a reduction/elimination in possible off-target effects, and (iii) precise control of the placement of the intended target sequence(s).
Sigma-Aldrich
氯化镁 六水合物, BioXtra, ≥99.0%
Sigma-Aldrich
L -精氨酸 单盐酸盐, reagent grade, ≥98% (HPLC), powder
Sigma-Aldrich
D -(+)-半乳糖, ≥99%
Sigma-Aldrich
L-谷氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-异亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
脱氧核糖核酸 钠盐 来源于鲑鱼睾丸
Sigma-Aldrich
L-天冬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-苏氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L -缬氨酸, reagent grade, ≥98% (HPLC)
Millipore
不含氨基酸和硫酸铵的酵母氮基础, suitable for microbiology, NutriSelect® Basic