Merck
  • 主页
  • 查找结果
  • [Optimization of Streptococcus equi subsp. zooepidemicus cultivation process--producer of hyaluronic acid].
[Optimization of Streptococcus equi subsp. zooepidemicus cultivation process--producer of hyaluronic acid].
Zhurnal mikrobiologii, epidemiologii, i immunobiologii (2013-06-29)
R N Tsepilov, A V Beloded, I I Samoĭlenko
摘要
Selection of high-mucoid morphotype of Streptococcus equi subsp. zooepidemicus (Streptococcus zooepidemicus) and study of its morphological, physiological, biochemical and technological characteristics for providing increased secretion of hyaluronic acid (HA). Submerged cultivation was performed in 100 ml glass flasks without baffles or in 1.5 or 10 1 laboratory bioreactors. LB and MRS media were used for cultivation. Mutagenesis was carried out by UV exposure with consequent selection of mucoid phenotype. HA was determined by carbazole method or after exhaustive acid hydrolysis by reaction of N-acetylglucosamine with Morgan-Elson reagent. Total hyaluronidase activity was evaluated by viscosimeter. Determination of cell and capsule size, ability to ferment carbohydrates and other microbiological, physiological and biochemical tests were performed by standard techniques. Instability of capsule phenotype of S. zooepidemicus B-8014 strain was revealed that is explained most probably by formation under certain conditions of bacterial hyaluronidase. This is confirmed by a reduction of HA concentration in cultural medium at pre- and stationary growth phases. Mucoid strain S. zooepidemicus KB-04 was obtained by mutagenesis with subsequent selection that is characterized by increased capsules. The strain was studied for HA formation. Optimization of growth medium composition, physical-chemical conditions and modes of cultivation allowed to significantly increase HA yield. The studies of morphologic, physiologic, biochemical and technological characteristics of the high-mucoid S. zooepidemicus KB-04 strain obtained by mutagenesis with consequent selection were performed, conditions of its cultivation and composition of growth mediu by carbon source and content of bivalent metal ions were optimized.
材料
货号
品牌
产品描述
Sigma-Aldrich
D -(+)-葡萄糖, ≥99.5% (GC)
Sigma-Aldrich
透明质酸酶 来源于牛睾丸, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
D -(+)-葡萄糖 溶液, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D -(+)-葡萄糖, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
葡萄糖, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D -(+)-葡萄糖, ACS reagent
Sigma-Aldrich
D -(+)-葡萄糖 溶液, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
葡萄糖, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D -(+)-葡萄糖, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
透明质酸酶 来源于牛睾丸, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
透明质酸 钠盐 来源于马链球菌, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
镁, turnings, reagent grade, 98%
Sigma-Aldrich
透明质酸酶 来源于绵羊睾丸, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
透明质酸酶 来源于牛睾丸, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
透明质酸 钠盐 来源于公鸡鸡冠, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
L - (−) -葡萄糖, ≥99%
Sigma-Aldrich
镁, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
透明质酸 钠盐 来源于牛玻璃体液
Sigma-Aldrich
D -(+)-葡萄糖, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D -(+)-葡萄糖, analytical standard
Sigma-Aldrich
透明质酸 钠盐 来源于马链球菌, mol wt 8,000-15,000
Sigma-Aldrich
透明质酸酶 来源于绵羊睾丸, Type II, lyophilized powder, ≥300 units/mg solid
透明质酸钠, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
透明质酸酶 来源于牛睾丸, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
D -(+)-葡萄糖, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
透明质酸 钠盐 来源于马链球菌, mol wt 1,000,000-1,250,000
Sigma-Aldrich
透明质酸 钠盐 来源于马链球菌, mol wt 1,500,000-1,750,000
Sigma-Aldrich
D -(+)-葡萄糖, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
透明质酸 钠盐 来源于兽疫链球菌, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
透明质酸 钠盐 来源于马链球菌, mol wt 2,000,000-2,400,000