  • Search Results
  • Characterization of disposable optical sensors for heavy metal determination.
Characterization of disposable optical sensors for heavy metal determination.
Talanta (2012-05-23)
Jadranka Vuković, María Ariza Avidad, Luis Fermín Capitán-Vallvey
ABSTRACT
This paper presents the development, characterization and quality control of analytical methods based on the use of disposable optical sensors for determination of heavy metals. Chromogenic reagents such as 1-(2-pyridylazo)-2-naphthol, (2-pyridylazo)resorcinol, Zincon, Ferrozine, and Chromazurol S were used to develop optical sensors of heavy metal ions found as contaminants in pharmaceutical substances and products, such as Zn(II), Cu(II), Ni(II), Fe(II), and Fe(III). The chromogenic reagents were immobilized in polymeric membranes by spin-coating from cocktails containing all reagents needed. The methods were prevalidated using a comprehensive quality control strategy based on a system of mathematical/statistical testing and diagnosis of each prevalidation step. This system involved characterization of analytical groups; checking of two limiting groups; testing of data homogeneity; recognition of outliers; and determination of analytical functions, limiting values, precision and accuracy. The prevalidation strategy demonstrated the reliability of the proposed method and pointed out some limitations. Combining the optical sensors with multicomponent linear regression allowed simultaneous determination of multiple metals in synthetic mixtures with different compositions. Good agreement between experimental and theoretical amounts of heavy metals in the mixtures was obtained for the majority of sensors and metals. Even better agreement was obtained between the experimental and theoretical total amounts of metals in the mixtures. The proposed analytical methods were successfully applied to the determination of zinc in pharmaceutical preparations of insulin and the determination of metal mixtures in a commercial nasal spray of isotonic seawater. The reliable and sensitive individual optical sensors developed in this study may be useful for designing a multimembrane optical tongue that with appropriate further optimization can be used for screening heavy metals in various matrices.
Sigma-Aldrich
3-(2-吡啶基)-5,6-二苯基-1,2,4-三嗪-4,4'-二磺酸 单钠盐 水合物, 97%
Sigma-Aldrich
铬天青 S, Dye content 50 %
Sigma-Aldrich
1-(2-吡啶偶氮)-2-萘酚, indicator grade
Supelco
3-(2-吡啶基)-5,6-二苯基-1,2,4-三嗪-4′,4′′-二磺酸 钠盐, for spectrophotometric det. of Fe, ≥97.0%
Sigma-Aldrich
3-(2-吡啶基)-5,6-二苯基-1,2,4-三嗪-4′,4′′-二磺酸 钠盐, Indicator ligand
Sigma-Aldrich
3-(2-吡啶基)-5,6-二苯基-1,2,4-三嗪-4′,4′′-二磺酸 钠盐, BioXtra