  • Search Results
  • Planar triazinium cations from vanadyl-mediated ring cyclizations: the thiazole species for efficient nuclear staining and photocytotoxicity.
Planar triazinium cations from vanadyl-mediated ring cyclizations: the thiazole species for efficient nuclear staining and photocytotoxicity.
Dalton transactions (Cambridge, England : 2003) (2013-01-31)
Puja Prasad, Imran Khan, Pijus K Sasmal, Debasis Koley, Paturu Kondaiah, Akhil R Chakravarty
ABSTRACT
Planar triazinium cationic species from vanadyl-assisted cyclization of 1-(2-thiazolylazo)-2-naphthol (H-TAN, 1), 1-(2-pyridylazo)-2-naphthol (H-PAN, 2), 2-(2'-thiazolylazo)-p-cresol (H-TAC, 3) and 6-(2'-thiazolylazo)-resorcinol (H-TAR, 5) were prepared and characterized. A dioxovanadium(V) species [VO(2)(TAR)] (4) was also isolated. Compounds 1, 2 and 4 were structurally characterized. Both 1 and 2 have planar structures. Complex 4 has V(V)O(3)N(2) coordination geometry. The cyclised triazinium compound forms a radical species within -0.06 to -0.29 V vs. SCE in DMF-0.1 M tetrabutylammonium perchlorate with a second response due to formation of an anionic species. A confocal microscopic study showed higher nuclear uptake for 1 having a fused thiazole moiety than 2 with a fused pyridine ring. The compounds showed a partial intercalative mode of binding to calf thymus DNA. Compound 1 showed plasmid DNA photo-cleavage activity under argon and photocytotoxicity in HeLa and MCF-7 cells with IC(50) values of 15.1 and 3.4 μM respectively in visible light of 400-700 nm, while being essentially non-toxic in the dark with IC(50) values of 90.4 and 21.9 μM. A TDDFT study was done to rationalize the experimental data.
Sigma-Aldrich
对甲酚, 99%
Sigma-Aldrich
脱氧核糖核酸 来源于小牛胸腺, Type XV, Activated, lyophilized powder
Sigma-Aldrich
1-(2-吡啶偶氮)-2-萘酚, indicator grade
Sigma-Aldrich
对甲酚, ≥99%, FG
Sigma-Aldrich
脱氧核糖核酸 来源于小牛胸腺, Genomic, unsheared
Supelco
对甲酚, analytical standard
Sigma-Aldrich
钒, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
脱氧核糖核酸,单链 来源于小牛胸腺, lyophilized powder
Sigma-Aldrich
对甲酚, puriss. p.a., ≥99.0% (GC)
Supelco
4-甲酚 溶液, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
钒, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
钒, rod, diam. 6.2 mm, 99.7% trace metals basis
Sigma-Aldrich
钒, turnings, 99.7% trace metals basis
Sigma-Aldrich
钒, foil, thickness 1.0 mm, 99.7%
Sigma-Aldrich
钒, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
钒, foil, thickness 0.5 mm, 99.7% trace metals basis