Merck
  • 主页
  • 查找结果
  • Purification of human immunoglobulin G autoantibodies to tumor necrosis factor using affinity chromatography and magnetic separation.
Purification of human immunoglobulin G autoantibodies to tumor necrosis factor using affinity chromatography and magnetic separation.
Journal of immunological methods (2013-02-08)
S V Sennikov, E A Golikova, F D Kireev, J A Lopatnikova
摘要
Autoantibodies to cytokines are important biological effector molecules that can regulate cytokine activities. The aim of the study was to develop a protocol to purify autoantibodies to tumor necrosis factor from human serum, for use as a calibration material to determine the absolute content of autoantibodies to tumor necrosis factor by enzyme-linked immunosorbent assay. The proposed protocol includes a set of affinity chromatography methods, namely, Bio-Gel P6DG sorbent to remove albumin from serum, Protein G Sepharose 4 Fast Flow to obtain a total immunoglobulin G fraction of serum immunoglobulins, and Affi-Gel 15 to obtain specifically antibodies to tumor necrosis factor. The addition of a magnetic separation procedure to the protocol eliminated contaminant tumor necrosis factor from the fraction of autoantibodies to tumor necrosis factor. The protocol generated a pure fraction of autoantibodies to tumor necrosis factor, and enabled us to determine the absolute concentrations of different subclasses of immunoglobulin G autoantibodies to tumor necrosis factor in apparently healthy donors.
材料
货号
品牌
产品描述
Sigma-Aldrich
琼脂糖, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
琼脂糖, Type I, low EEO
Sigma-Aldrich
琼脂糖, for molecular biology
Sigma-Aldrich
琼脂糖, Low EEO
Sigma-Aldrich
琼脂糖, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
琼脂糖, Medium EEO, for molecular biology
Sigma-Aldrich
Sepharose® 4B, 45-165 μm bead diameter
Sigma-Aldrich
琼脂糖, Type I-A, low EEO
Sigma-Aldrich
琼脂糖, Ultra-low Gelling Temperature
Sigma-Aldrich
琼脂糖, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
琼脂糖, Type II-A, Medium EEO
Sigma-Aldrich
琼脂糖, High Gelling Temperature
Sigma-Aldrich
琼脂糖, High EEO, for molecular biology
Sigma-Aldrich
Sepharose® 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
琼脂糖, Wide range, for molecular biology
Sigma-Aldrich
琼脂糖, High EEO
Sigma-Aldrich
琼脂糖, For pulsed field electrophoresis running gel
Sigma-Aldrich
琼脂糖, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
琼脂糖, Special High EEO
Sigma-Aldrich
琼脂糖, Type IV, Special High EEO
Sigma-Aldrich
琼脂糖, Vetec, reagent grade