  • Search Results
  • Combustion of volatile organic compounds over composite catalyst of Pt/γ-Al₂O₃ and beta zeolite.
Combustion of volatile organic compounds over composite catalyst of Pt/γ-Al₂O₃ and beta zeolite.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2013-03-01)
Yasuyuki Takamitsu, Satoshi Yoshida, Wataru Kobayashi, Hiroshi Ogawa, Tsuneji Sano
ABSTRACT
Catalytic oxidation of volatile organic compounds (VOCs) was carried out over a composite catalyst comprising Pt/γ-Al₂O₃ and protonated beta zeolite. The conversion of several VOCs such as ethyl acetate, butyl acetate, 2-propanol, 1,2-dichloroethane, and chloroethane over the composite catalyst was higher than the conversion over the conventional Pt/γ-Al₂O₃ catalyst, indicating a remarkable improvement in the oxidation activity of the composite. On the other hand, no difference in the conversion of methyl ethyl ketone, methyl isobutyl ketone, toluene, benzene, and chloroethylene was observed for the composite catalyst versus the Pt/γ-Al₂O₃ catalyst. To clarify the role of the zeolite component, the reaction products obtained using the composite catalyst were compared with those obtained using the Pt/γ-Al₂O₃ catalyst. For the cases in which considerable improvement in the oxidation activity was observed with the composite, it was revealed that the conversion of VOCs to intermediate compounds took place over the acidic sites of the zeolite; the intermediates tended to be easily oxidized to CO₂ on the Pt/γ-Al₂O₃ catalyst. In addition, the composite catalyst also exhibited high durability. High catalytic activity was maintained even after aging at 600°C for more than 1000 h.
Sigma-Aldrich
沸石
Sigma-Aldrich
介孔结构的硅铝酸盐, MCM-41 (hexagonal)