  • Search Results
  • A boron dipyrromethene-phthalocyanine pentad as an artificial photosynthetic model.
A boron dipyrromethene-phthalocyanine pentad as an artificial photosynthetic model.
Chemical communications (Cambridge, England) (2013-03-06)
Jian-Yong Liu, Yingsi Huang, Roel Menting, Beate Röder, Eugeny A Ermilov, Dennis K P Ng
ABSTRACT
A silicon(IV) phthalocyanine with two axial p-phenylene-linked boron dipyrromethene and monostyryl boron dipyrromethene moieties has been prepared. The resulting pentad absorbs strongly in most of the UV-visible region and serves as an artificial photosynthetic antenna-reaction centre model.
Sigma-Aldrich
硼, crystalline, 1 cm, 99.7% trace metals basis
Sigma-Aldrich
紫质胆素原, powder