  • Search Results
  • [Ionic conjugates of sulfate carboxymethyl cellulose with dialkylaminomethyl derivatives of 2-isobornyl-4-methylphenol: synthesis and study of anti-inflammatory and analgesic activity].
[Ionic conjugates of sulfate carboxymethyl cellulose with dialkylaminomethyl derivatives of 2-isobornyl-4-methylphenol: synthesis and study of anti-inflammatory and analgesic activity].
Bioorganicheskaia khimiia (2013-04-04)
I Iu Chukicheva, E V Buravlev, M A Torlopov, Iu B Vikharev, L V Anikina, V V Grishko
ABSTRACT
Water-soluble sulfate polysalts of carboxymethyl cellulose and tertiary aminomethyl derivatives of 2-isobornyl-4-methylphenol was obtained. For the synthesized conjugates investigated in vivo anti-inflammatory activity in the test of acute formalin inflammation and analgesic activity in tests the "hot plate" and "vinegar cramps".
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
羧甲基纤维素钠, average Mw ~90,000
Sigma-Aldrich
羧甲基纤维素 钠盐, low viscosity
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
甲醛 溶液, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
羧甲基纤维素 钠盐, Medium viscosity
Sigma-Aldrich
羧甲基纤维素 钠盐, High viscosity
Sigma-Aldrich
羧甲基纤维素钠, average Mw ~700,000
Sigma-Aldrich
甲醛 溶液, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
羧甲纤维素 钠, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
羧甲基纤维素钠, average Mw ~250,000, degree of substitution 0.7
Supelco
甲醛 溶液, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
羧甲基纤维素钠, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
羧甲基纤维素钠, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
甲醛 溶液, tested according to Ph. Eur.
Sigma-Aldrich
羧甲基纤维素 钠盐, medium viscosity
Sigma-Aldrich
羧甲基纤维素 钠盐, ultra high viscosity, highly purified