Merck
Long-term cytotoxic effects of contemporary root canal sealers.
Journal of applied oral science : revista FOB (2013-04-06)
Emmanuel João Nogueira Leal da Silva, Carolina Carvalho Santos, Alexandre Augusto Zaia
摘要
The aim of the present study was to investigate the effects of root canal sealers on the cytotoxicity of 3T3 fibroblasts during a period of 5 weeks. Fibroblasts (3T3, 1×105 cells per well) were incubated with elutes of fresh specimens from eight root canal sealers (AH Plus, Epiphany, Endomethasone N, EndoREZ, MTA Fillapex, Pulp Canal Sealer EWT, RoekoSeal and Sealapex) and with elutes of the same specimens for 5 succeeding weeks after immersing in simulated body fluid. The cytotoxicity of all root canal sealers was determined using the MTT assay. Data were analyzed using ANOVA and Tukey's test. RoekoSeal was the only sealer that did not show any cytotoxic effects (p<0.05). All the other tested sealers exhibited severe toxicity initially (week 0). MTA Fillapex remained moderately cytotoxic after the end of experimental period. Toxicity of the other tested sealers decreased gradually over time. The evaluated root canal sealers presented varying degrees of cytotoxicity, mainly in fresh mode. RoekoSeal had no cytotoxic effect both freshly mixed and in the other tested time points. MTA Fillapex was associated with significantly less cell viability when compared to the other tested root canal sealers.
材料
货号
品牌
产品描述
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
氢氧化钙, ACS reagent, ≥95.0%
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
麝香草酚, ≥98.5%
SAFC
甲醛 溶液, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
甲醛 溶液, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
麝香草酚, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Supelco
甲醛 溶液, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
麝香草酚, FCC, FG
Sigma-Aldrich
甲醛 溶液, tested according to Ph. Eur.
Supelco
麝香草酚, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
氢氧化钙, 99.995% trace metals basis
Sigma-Aldrich
氢氧化钙, puriss. p.a., reag. Ph. Eur., ≥96%
Supelco
麝香草酚, analytical standard
麝香草酚, European Pharmacopoeia (EP) Reference Standard
麝香草酚, primary reference standard