  • Search Results
  • Adaptive appetites for salted and unsalted food in rats: differential effects of sodium depletion, DOCA, and dehydration.
Adaptive appetites for salted and unsalted food in rats: differential effects of sodium depletion, DOCA, and dehydration.
American journal of physiology. Regulatory, integrative and comparative physiology (2013-04-19)
M J McKinley
ABSTRACT
Most ingested sodium is contained in food. The aim was to investigate whether sodium depletion, dehydration, or DOCA alters intakes of salted and unsalted foods by rats given choices of two foods: salted (0.2-0.5% Na) and unsalted food containing either similar or different other dietary components. Diuretic-induced (furosemide or acetazolamide, two treatments on successive days) sodium depletion always caused pronounced falls in intake of unsalted food within 24 h, continuing at least another 2 days (e.g., 20.9 ± 1.6 pretreatment to 14.8 ± 1.2, 10.6 ± 1.5, and 14.3 ± 1.3 g/day for 3 days of depletion). Intake and preference for salted food increased after 24-72 h (e.g., 6.5 ± 1.2 pretreatment to 7.1 ± 1.1, 16.4 ± 2.3, and 17.0 ± 1.5 g/day at 1, 2, and 3 days of depletion). Valsartan (10 mg/day) blocked the increased intake of salted food but not the reduced intake of unsalted food. DOCA (2 mg/day) caused equivalent increase and decrease in intakes of salted and unsalted food, respectively. Water-deprived rats reduced intake (e.g., 14.2 ± 3.1 to 3.2 ± 2.0 g/day) of and preference for salted food (e.g., 56 ± 13% to 21 ± 11%) after 2 days of dehydration but did not consistently reduce intake of unsalted food. Total food ingested/day fell in both sodium-depleted and dehydrated rats. Rats regulate intakes of different foods to balance sodium needs, osmoregulatory homeostasis, and energy requirements. Reduced appetite for unsalted food may be a homeostatic response to sodium depletion, which together with subsequent generation of appetite for salted food, drives animals to ingest sodium-containing food, thereby restoring sodium balance.
Sigma-Aldrich
L -缬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L -缬氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
呋塞米
SAFC
L -缬氨酸
Supelco
呋塞米, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
乙酰唑胺, ≥99%, powder
Supelco
L-缬氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
缬沙坦, ≥98% (HPLC)
Sigma-Aldrich
L -缬氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
呋塞米, European Pharmacopoeia (EP) Reference Standard
Supelco
L -缬氨酸, certified reference material, TraceCERT®
缬沙坦, European Pharmacopoeia (EP) Reference Standard
Supelco
呋塞米 溶液, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
乙酰唑胺, European Pharmacopoeia (EP) Reference Standard
缬沙坦, European Pharmacopoeia (EP) Reference Standard