Merck
  • 主页
  • 查找结果
  • The mechanism by which a propeptide-encoded pH sensor regulates spatiotemporal activation of furin.
The mechanism by which a propeptide-encoded pH sensor regulates spatiotemporal activation of furin.
The Journal of biological chemistry (2013-05-09)
Danielle M Williamson, Johannes Elferich, Parvathy Ramakrishnan, Gary Thomas, Ujwal Shinde
摘要
The proprotein convertase furin requires the pH gradient of the secretory pathway to regulate its multistep, compartment-specific autocatalytic activation. Although His-69 within the furin prodomain serves as the pH sensor that detects transport of the propeptide-enzyme complex to the trans-Golgi network, where it promotes cleavage and release of the inhibitory propeptide, a mechanistic understanding of how His-69 protonation mediates furin activation remains unclear. Here we employ biophysical, biochemical, and computational approaches to elucidate the mechanism underlying the pH-dependent activation of furin. Structural analyses and binding experiments comparing the wild-type furin propeptide with a nonprotonatable His-69 → Leu mutant that blocks furin activation in vivo revealed protonation of His-69 reduces both the thermodynamic stability of the propeptide as well as its affinity for furin at pH 6.0. Structural modeling combined with mathematical modeling and molecular dynamic simulations suggested that His-69 does not directly contribute to the propeptide-enzyme interface but, rather, triggers movement of a loop region in the propeptide that modulates access to the cleavage site and, thus, allows for the tight pH regulation of furin activation. Our work establishes a mechanism by which His-69 functions as a pH sensor that regulates compartment-specific furin activation and provides insights into how other convertases and proteases may regulate their precise spatiotemporal activation.
材料
货号
品牌
产品描述
Sigma-Aldrich
甘油, for molecular biology, ≥99.0%
Sigma-Aldrich
甘油, ≥99.5%
Sigma-Aldrich
尿素, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
甘油, ACS reagent, ≥99.5%
Sigma-Aldrich
尿素, ACS reagent, 99.0-100.5%
Sigma-Aldrich
甘油, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
甘油, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
L-组氨酸, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Supelco
甘油, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
尿素, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
甘油, meets USP testing specifications
Sigma-Aldrich
甘油 溶液, 83.5-89.5% (T)
Sigma-Aldrich
尿素 溶液, BioUltra, ~8 M in H2O
Supelco
尿素, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
甘油, BioXtra, ≥99% (GC)
SAFC
L-组氨酸
Sigma-Aldrich
尿素, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
尿素, suitable for electrophoresis
Sigma-Aldrich
甘油, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
甘油, FCC, FG
Sigma-Aldrich
L-组氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
尿素, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
尿素, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
尿素, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
甘油, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
甘油, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
甘油, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
尿素 溶液, 40 % (w/v) in H2O
Sigma-Aldrich
甘油 溶液, puriss., meets analytical specification of Ph. Eur., BP, 84-88%