Merck
  • 主页
  • 查找结果
  • The effects of liver impairment on the pharmacokinetics of brivanib, a dual inhibitor of fibroblast growth factor receptor and vascular endothelial growth factor receptor tyrosine kinases.
The effects of liver impairment on the pharmacokinetics of brivanib, a dual inhibitor of fibroblast growth factor receptor and vascular endothelial growth factor receptor tyrosine kinases.
Cancer chemotherapy and pharmacology (2013-05-31)
Anthony El-Khoueiry, James A Posey, Juan Ramón Castillo Ferrando, Smitha S Krishnamurthi, Shariq Syed, Georgia Kollia, Ian Walters, Bruce S Fischer, Eric Masson
摘要
Hepatic impairment may impede tyrosine kinase inhibitor metabolism. This phase I study compared the pharmacokinetics of brivanib in patients with hepatocellular carcinoma (HCC) and varying levels of hepatic impairment with those with non-HCC malignancies and normal liver function. Patients were assigned to the following groups: Groups A, B, and C (HCC plus mild, moderate, or severe hepatic impairment, respectively) and Group D (non-HCC malignancy and normal hepatic function). Brivanib alaninate (brivanib prodrug) doses were 400 mg in Groups A, B, and D and 200 mg in Group C. Brivanib exposure was determined on day 1 (single dose) and day 28 (multiple doses). Twenty-four patients participated in the study. After a single brivanib alaninate dose, brivanib exposure was comparable between Groups A, B, and D. Area under the concentration-time curve was 50 % higher in Group C versus Group D. There were not enough data to draw conclusions on multiple doses. Safety profile in Groups A, B, and D was consistent with previous brivanib monotherapy experience. Tolerability could not be assessed in Group C because of dose interruptions and discontinuations, generally due to the disease natural history. Brivanib exposure was similar in patients with HCC and mild or moderate hepatic impairment (Child-Pugh [CP] A or B status) and those with non-HCC malignancies and normal hepatic function, suggesting dose adjustment is unnecessary with CP A or B status. Experience with HCC and severe hepatic impairment (CP C status) is insufficient to recommend brivanib use in this population.
材料
货号
品牌
产品描述
Sigma-Aldrich
L-丙氨酸, ≥98% (TLC)
Sigma-Aldrich
L-丙氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-丙氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-丙氨酸, ≥99%
Supelco
L-丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-丙氨酸, ≥99.0% (NT)
丙氨酸, European Pharmacopoeia (EP) Reference Standard
Supelco
L-丙氨酸, certified reference material, TraceCERT®
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C