  • Search Results
  • Structural definition is important for the propagation of the yeast [PSI+] prion.
Structural definition is important for the propagation of the yeast [PSI+] prion.
Molecular cell (2013-06-12)
Ricardo Marchante, Michelle Rowe, Jo Zenthon, Mark J Howard, Mick F Tuite
ABSTRACT
Prions are propagated in Saccharomyces cerevisiae with remarkable efficiency, yet we know little about the structural basis of sequence variations in the prion protein that support or prohibit propagation of the prion conformation. We show that certain single-amino-acid substitutions in the prion protein Sup35 impact negatively on the maintenance of the associated prion-based [PSI(+)] trait by combining in vivo phenotypic analysis with solution NMR structural studies. A clear correlation is observed between mutationally induced conformational differences in one of the oligopeptide repeats (R2) in the N terminus of Sup35 and the relative ability to propagate [PSI(+)]. Strikingly, substitution of one of a Gly-Gly pair with highly charged residues that significantly increase structural definition of R2 lead to a severe [PSI(+)] propagation defect. These findings offer a molecular explanation for the dominant-negative effects of such psi-no-more (PNM) mutations and demonstrate directly the importance of localized structural definition in prion propagation.
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
SAFC
L-酪氨酸
Sigma-Aldrich
L-酪氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-酪氨酸, BioUltra, ≥99.0% (NT)
Supelco
L-酪氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-酪氨酸, FG
Supelco
L-酪氨酸, certified reference material, TraceCERT®