Merck
  • 主页
  • 查找结果
  • Preliminary studies on palladium nanoparticle as a novel label for DNA microarray and their corresponding detection.
Preliminary studies on palladium nanoparticle as a novel label for DNA microarray and their corresponding detection.
Journal of biomedical nanotechnology (2013-07-19)
Zhifei Wang, Hongyin Li, Shuang Zhen, Yuanying Zhang, Nongyue He
摘要
This paper firstly describes the preliminary results achieved by using palladium nanoparticle (Pd NP) as a novel label for the detection of DNA hybridization in DNA microarray. And two signal amplification procedures based on "the silver staining" or "the cobalt staining" are presented during above analysis. The results show that the label Pd NP-ssDNA (target) (single strand DNA(target)) performs high single base pair mismatch-discrimination capability. The succeeding silver staining or cobalt staining procedure greatly amplifies such a signal through the catalysis of Pd. For "the silver staining:' the background staining is very low and the silver deposition only occurs around Pd NPs. So such a procedure provides a alternative for "Gold Label Silver Stain" presented by Mirkin C. A. For "the cobalt staining," not only a colorimetric array but also a magnetic sensor (such as Magnetic Tunnel Junction sensor, MTJ) can be used to detect the obtained cobalt dot due to its strong magnetic property, which provides a new strategy for DNA microarray detection. So as the proof-of-concept investigations, this work proved the feasibility of the application of Pd NPs as the label in DNA microarray assay.
材料
货号
品牌
产品描述
Sigma-Aldrich
脱氧核糖核酸,单链 来源于鲑鱼睾丸, For hybridization
Sigma-Aldrich
脱氧核糖核酸,单链 来源于鲑鱼睾丸, For hybridization
Sigma-Aldrich
钯, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
脱氧核糖核酸 钠盐 来源于鲱鱼睾丸, Type XIV
Sigma-Aldrich
钯, powder, 99.995% trace metals basis
Sigma-Aldrich
质粒DNA 来源于大肠杆菌 RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
钯, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
钯黑, surface area 40-60 m2/g, 99.95% trace metals basis
Sigma-Aldrich
脱氧核糖核酸 钠盐 来源于人类胎盘, Type XIII
Sigma-Aldrich
钯, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
脱氧核糖核酸 来源于人类胎盘, buffered aqueous solution, sexed, female
Sigma-Aldrich
钯, granular, 99.99% trace metals basis
Sigma-Aldrich
钯, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
钯, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
钯, foil, thickness 0.05 mm, 99.9% trace metals basis
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from human placenta
Sigma-Aldrich
钯, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
钯, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
钯, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
钯, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
钯, sponge, 99.9% trace metals basis
Sigma-Aldrich
钯, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis