• 主页
  • 查找结果
  • Methane catalytic combustion on Pd9/gamma-Al2O3 with different degrees of Pd oxidation.
Methane catalytic combustion on Pd9/gamma-Al2O3 with different degrees of Pd oxidation.
Chimia (2013-08-24)
Izabela Czekaj, Katarzyna A Kacprzak, John Mantzaras
摘要
This research is focused on the analysis of adsorbed CH4 intermediates at oxidized Pd9 nanoparticles supported on gamma-alumina. From first-principle density functional theory (DFT) calculations, several configurations, charge transfer and electronic density of states have been analyzed in order to determine feasible paths for the oxidation process. Furthermore methane oxidation cycles have been investigated on Pd nanoparticles with different degrees of oxidation. In case of low oxidized Pd nanoparticles, activation of methane is observed, whereby hydrogen from methane is adsorbed at one oxygen atom. This reaction is exothermic. In a subsequent step, two water molecules desorb. Additionally, a very interesting structural effect becomes evident; Pd-carbide formation, which is also an exothermic reaction. Furthermore, oxidation of such carbidized Pd-nanoparticles leads to CO2 formation, which is an endothermic reaction. One important result is that the support is involved in the CO2 formation. A different mechanism of methane oxidation was discovered for highly oxidized Pd nanoparticles. When the Pd nanoparticle is more strongly exposed to oxidative conditions, adsorption of methane is also possible, but it leads to carbonic acid production at the interface between the Pd nanoparticles and support. This process is endothermic.
材料
货号
品牌
产品描述
Sigma-Aldrich
氧化铝, activated, basic, Brockmann I
Sigma-Aldrich
氧化铝, activated, neutral, Brockmann I
Sigma-Aldrich
氧化铝, nanopowder, <50 nm particle size (TEM)
Supelco
氧化铝, activated, neutral, Brockmann Activity I
Sigma-Aldrich
氧化铝, powder, 99.99% trace metals basis
Sigma-Aldrich
氧化铝, activated, acidic, Brockmann I
Sigma-Aldrich
氧化铝, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
氧化铝, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
钯, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
氧化铝, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
氧化铝, 99.997% trace metals basis
Sigma-Aldrich
氧化铝, pellets, 3 mm
Sigma-Aldrich
氧化铝, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
钯, powder, 99.995% trace metals basis
Sigma-Aldrich
钯黑, surface area 40-60 m2/g, 99.95% trace metals basis
Sigma-Aldrich
氧化铝, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
氧化铝, Corundum, α-phase, -100 mesh
Sigma-Aldrich
介孔氧化铝, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
氧化铝, pore size 58 Å, ~150 mesh
Sigma-Aldrich
氧化铝, single crystal substrate, <0001>
Sigma-Aldrich
钯, granular, 99.99% trace metals basis
Sigma-Aldrich
钯, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
氧化铝, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
氧化铝, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
氧化铝, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
钯, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
钯, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
钯, sponge, 99.9% trace metals basis
Supelco
氧化铝, for the determination of hydrocarbons
Sigma-Aldrich
氧化铝, fused, powder, primarily α-phase, 100-200 mesh