Merck
  • 主页
  • 查找结果
  • High concentrations of the melatonin metabolite, N1-acetyl-N2-formyl-5-methoxykynuramine, in cerebrospinal fluid of patients with meningitis: a possible immunomodulatory mechanism.
High concentrations of the melatonin metabolite, N1-acetyl-N2-formyl-5-methoxykynuramine, in cerebrospinal fluid of patients with meningitis: a possible immunomodulatory mechanism.
Journal of pineal research (2005-09-10)
Sueli de Oliveira Silva, Valdecir F Ximenes, José A Livramento, Luiz H Catalani, Ana Campa
摘要
We evaluated the presence of the melatonin metabolite N1-acetyl-N2-formyl-5-methoxykynuramine (AFMK), in cerebrospinal fluid (CSF) of patients with viral meningitis (n = 20) and control samples (n = 8) and correlate AFMK levels with inflammatory markers such as cellularity, protein, tumor necrosis factor (TNF)-alpha, interleukin (IL)-8 and IL-1beta levels. A portion of the CSF was extracted with dichloromethane (1:5) and analyzed by high-performance liquid chromatography (HPLC) under standardized conditions for AFMK. AFMK was detected in 16 of 20 CSF samples of patients with viral meningitis; the concentration of AFMK was found to be above the quantification limit (50 nmol/L) in six of these samples. AFMK was not detected in any of the eight control samples. The samples were classified into groups according to AFMK levels: undetectable (<10 nmol/L, group I), detectable but below the quantification limit (< 50 nmol/L, group II), and quantified (>50 nmol/L, group III). Group II presented the highest levels of proteins and IL-8, whereas group III showed the lowest levels of the inflammatory parameters. This study supports our hypothesis that inflammation favors the formation of AFMK and that this compound has immunomodulatory activity in vivo.
材料
货号
品牌
产品描述
Sigma-Aldrich
尿素, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
尿素, ACS reagent, 99.0-100.5%
Sigma-Aldrich
尿素, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
丙酮酸, 98%
Sigma-Aldrich
γ-氨基丁酸, ≥99%
Sigma-Aldrich
尿素 溶液, BioUltra, ~8 M in H2O
Sigma-Aldrich
L-丙氨酸, ≥98% (TLC)
Supelco
尿素, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
尿素, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
尿素, suitable for electrophoresis
Sigma-Aldrich
尿素, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
腺苷-3′,5′-环单磷酸, ≥98.5% (HPLC), powder
Sigma-Aldrich
尿素, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
L-丙氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-丙氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
尿素, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
丙酮酸, ≥97%, FG
Sigma-Aldrich
(R)-3-羟基丁酸, ≥98.0% (T)
Sigma-Aldrich
L-丙氨酸, ≥99%
Sigma-Aldrich
尿素 溶液, 40 % (w/v) in H2O
Sigma-Aldrich
γ-氨基丁酸, BioXtra, ≥99%
Sigma-Aldrich
尿素, meets USP testing specifications
Supelco
L-丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
丙酮酸, natural, ≥95%, FG
Millipore
尿素 溶液, suitable for microbiology, 40% in H2O
Sigma-Aldrich
L-丙氨酸, ≥99.0% (NT)
Sigma-Aldrich
丙酮酸, 95%
丙氨酸, European Pharmacopoeia (EP) Reference Standard
Supelco
L-丙氨酸, certified reference material, TraceCERT®
尿素, European Pharmacopoeia (EP) Reference Standard