  • Search Results
  • Plasma free amino acid concentrations in healthy Guatemalan adults and in patients with classic dengue.
Plasma free amino acid concentrations in healthy Guatemalan adults and in patients with classic dengue.
The American journal of clinical nutrition (2001-03-10)
P Klassen, P Fürst, C Schulz, M Mazariegos, N W Solomons
ABSTRACT
Plasma free amino acid patterns in health and disease have been reported. However, amino acid concentrations in adult populations in developing countries and in patients with dengue, as a model for an acute infectious viral disease endemic to the tropics, have not been reported. The purpose of this study was to determine the amino acid profile in both healthy Guatemalan adults from different socioeconomic backgrounds and at 3 time points during the course of classic dengue. The study was carried out in Guatemala and included measurement of plasma free amino acids in 22 healthy control subjects (14 low income, 8 middle class) and 17 febrile patients. Measurements of amino acids were repeated within a 48-h interval in 20 of the healthy Guatemalans. In 9 patients with dengue, amino acids were assayed 3 times: on admission to a local hospital in the coastal plain of Guatemala, on hospital discharge, and 7 d after hospital discharge. Branched-chain amino acid concentrations in healthy adults and dengue patients in Guatemala were lower than normal values reported in the literature for healthy Swedish adults. With the exception of increased phenylalanine concentrations and an increased ratio of phenylalanine to tyrosine, all amino acids as well as the Fischer molar ratio were decreased in the acute phase of dengue. Healthy Guatemalans have different amino acid patterns than do Swedish subjects independent of socioeconomic status. The systemic viral disease dengue is associated with changes in the plasma free amino acid pattern, reflecting infection-related alterations in amino acid metabolism.
Sigma-Aldrich
尿素, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
尿素, ACS reagent, 99.0-100.5%
Sigma-Aldrich
尿素, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Supelco
尿素, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
L-丙氨酸, ≥98% (TLC)
Sigma-Aldrich
尿素, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
尿素 溶液, BioUltra, ~8 M in H2O
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
尿素, suitable for electrophoresis
Sigma-Aldrich
L-丙氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
尿素, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
尿素, BioUltra, for molecular biology, ≥99% (T)
Supelco
L-苯基丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
尿素, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
L-丙氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
尿素, meets USP testing specifications
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Supelco
L-丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-丙氨酸, ≥99%
Sigma-Aldrich
L-苯基丙氨酸, 99%
Sigma-Aldrich
L-丙氨酸, ≥99.0% (NT)
Sigma-Aldrich
尿素 溶液, 40 % (w/v) in H2O
Millipore
尿素 溶液, suitable for microbiology, 40% in H2O
Supelco
L-苯基丙氨酸, certified reference material, TraceCERT®
丙氨酸, European Pharmacopoeia (EP) Reference Standard
Supelco
L-苯基丙氨酸, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Supelco
L-丙氨酸, certified reference material, TraceCERT®
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC
尿素, European Pharmacopoeia (EP) Reference Standard