  • Search Results
  • High-performance liquid chromatographic determination of acetone in blood and urine in the clinical diagnostic laboratory.
High-performance liquid chromatographic determination of acetone in blood and urine in the clinical diagnostic laboratory.
Journal of chromatography (1991-08-16)
A Brega, P Villa, G Quadrini, A Quadri, C Lucarelli
ABSTRACT
A method for the determination of acetone in plasma or urine by high-performance liquid chromatography (HPLC) was developed. Plasma specimens are deproteinized with acetonitrile (1:1, v/v) 2,4-dinitrophenylhydrazine (DNPH) is added to the supernatant or to filtered urine samples, similarly treated with acetonitrile (2:1, v/v) to prevent crystallization of the synthesized phenylhydrazone. An aliquot (20 microliters) of the reaction mixture was subjected to HPLC at ambient temperature using a reversed-phase Pecosphere 3 x 3 C18 column with acetonitrile-water (45:55, v/v) as eluent at a flow-rate of 1 ml/min and detection at 365 nm. Hydroxyacetone and acetoacetate phenylhydrazone derivatives do not interfere. The identification of acetone by its retention time was confirmed by comparison with a laboratory-synthesized acetone DNPH derivative. The concentration of acetone, eluted within 3 min, was determined by the peak-height method. The detection limit was 0.034 mmol/l; the relative standard deviations were less than 5% within run (n = 20) and less than 10% between run (n = 20).
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
丙酮, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
丙酮, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.8%
Sigma-Aldrich
丙酮, Laboratory Reagent, ≥99.5%
Supelco
丙酮, analytical standard
Supelco
丙酮, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
丙酮, ACS reagent, ≥99.5%
Sigma-Aldrich
丙酮, suitable for HPLC, ≥99.9%
Sigma-Aldrich
丙酮, histological grade, ≥99.5%
Sigma-Aldrich
丙酮, natural, ≥97%
Sigma-Aldrich
丙酮, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
丙酮 溶液, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
丙酮, ≥99%, meets FCC analytical specifications