  • Search Results
  • Quantification of neurotransmitter amino acids by capillary electrophoresis laser-induced fluorescence detection in biological fluids.
Quantification of neurotransmitter amino acids by capillary electrophoresis laser-induced fluorescence detection in biological fluids.
Analytical and bioanalytical chemistry (2010-08-31)
Angelo Zinellu, Salvatore Sotgia, Elisabetta Pisanu, Bastianina Scanu, Manuela Sanna, Maria Franca Usai, Roberto Chessa, Luca Deiana, Ciriaco Carru
ABSTRACT
The role of neurotransmitter amino acids (NAAs) in the functioning of the nervous system has been the focus of increasingly intense research over the past several years. Among the various amino acids that have important roles as neurotransmitters, there are alanine (Ala), glutamic acid (Glu), aspartic acid (Asp), serine (Ser), taurine (Tau) and glycine (Gly). NAAs are present in plasma, cells and--at trace levels--in all biological fluids, but complex components in biological matrices make it difficult to determine them in biological samples. We describe a new capillary electrophoresis (CE) method with laser-induced fluorescence detection by which analytes are resolved in less than 12 minutes in a 18 mmol/L phosphate run buffer at pH 11.6. The use of elevated temperatures during sample derivatization leads to a drastic reduction in the reaction time, down to 20 min, compared to the 6-14 h usually described for reactions between FITC and amino acids at room temperature. In order to demonstrate its wide range of applications, the method was applied to the analysis of NAA in human plasma and in other sample types, such as red blood cells, urine, cultured cells, cerebrospinal fluid, saliva and vitreous humor, thus avoiding the typical limitations of other methods, which are normally suitable for use with only one or two matrix types.
Sigma-Aldrich
甘氨酸, suitable for electrophoresis, ≥99%
Sigma-Aldrich
甘氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
甘氨酸, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
甘氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
甘氨酸, ACS reagent, ≥98.5%
SAFC
甘氨酸
Sigma-Aldrich
甘氨酸, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
甘氨酸, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
甘氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
牛磺酸, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
1 M 甘氨酸 溶液
Supelco
牛磺酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
甘氨酸, BioXtra, ≥99% (titration)
Sigma-Aldrich
牛磺酸, BioUltra, ≥99.5% (T)
Supelco
甘氨酸, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
甘氨酸, 99%, FCC
SAFC
牛磺酸
Sigma-Aldrich
牛磺酸, ≥98%, FG
Supelco
甘氨酸, certified reference material, TraceCERT®
Sigma-Aldrich
甘氨酸, tested according to Ph. Eur.