  • Search Results
  • Examination of urine metabolites in the newborn period and during protein loading tests at 6 months of age--Part 1.
Examination of urine metabolites in the newborn period and during protein loading tests at 6 months of age--Part 1.
European journal of pediatrics (1990-01-01)
E Mönch, J Kneer, C Jakobs, M Arnold, H Diehl, U Batzler
ABSTRACT
In the course of the collaborative study of children treated for PKU, urine samples from a total of 165 patients were analysed at six different times: in the newborn period before onset of therapy, after beginning of dietary management, during and immediately after a protein loading test at 6 months of age. In 95.9% of newborns with elevated Phe levels in plasma, metabolites of this amino acid as well as of Tyr could be detected. Of all metabolites phenylpyruvate always showed the highest concentration, followed by phenyllactate and o-hydroxy-phenylacetate. During the protein loading test an increase of the same metabolites occurred. At the age of 6 months the percentage of p-hydroxylated compounds related to the sum of all metabolites was lower than in the newborn period. Comparing the results of urine analyses at 6 months of age after the protein loading tests with the classification of HPA into the reaction types I-III, it can be clearly stated that patients with the milder forms II and III have already lower levels of Phe metabolites in urine before onset of therapy compared to the reaction type I. In retrospect 52% of the newborns could therefore be classified as reaction type I even before beginning of dietary management. The analysis of urinary Phe metabolites before the onset of therapy therefore provides sufficient information about the reaction type.
Sigma-Aldrich
L-亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-亮氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Sigma-Aldrich
L -缬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L -缬氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
苯乙酸, 99%
Sigma-Aldrich
扁桃酸, 99%
SAFC
L -缬氨酸
Sigma-Aldrich
L-亮氨酸, BioUltra, ≥99.5% (NT)
Supelco
L-苯基丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-亮氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
苯乙酸, ≥99%, FCC, FG
Supelco
L-缬氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-亮氨酸, 99%, FG
Sigma-Aldrich
苯丙酮酸, 98%
Sigma-Aldrich
3-苯基乳酸, ≥98%
Sigma-Aldrich
17α-羟基孕酮, ≥95%
Sigma-Aldrich
L-苯基丙氨酸, 99%
Sigma-Aldrich
L -缬氨酸, BioUltra, ≥99.5% (NT)
Supelco
17α-羟孕酮标准液 溶液, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
DL-扁桃酸, analytical standard
Sigma-Aldrich
邻羟基苯乙酸, ReagentPlus®, 99%
Sigma-Aldrich
L-(-)-3-苯基乳酸, 98%
Sigma-Aldrich
3-羟基苯乙酸, ≥99%
Supelco
L-苯基丙氨酸, certified reference material, TraceCERT®
Supelco
L-苯基丙氨酸, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Supelco
L-亮氨酸, certified reference material, TraceCERT®
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC