  • Search Results
  • Amino acids in plasma and CSF and monoamine metabolites in CSF: interrelationship in healthy subjects.
Amino acids in plasma and CSF and monoamine metabolites in CSF: interrelationship in healthy subjects.
Journal of neurochemistry (1984-03-01)
L Hagenfeldt, L Bjerkenstedt, G Edman, G Sedvall, F A Wiesel
ABSTRACT
Plasma and cerebrospinal fluid (CSF) concentrations of amino acids were measured in 65 healthy volunteers (50 men and 15 women). The CSF levels of the monoamine metabolites homovanillic acid (HVA), 3-methoxy-4-hydroxyphenylethylene glycol (MOPEG), and 5-hydroxyindoleacetic acid (5-HIAA) were also determined. Sex differences were observed in both plasma and CSF amino acid levels as well as in the relationship between these concentrations. No significant correlations were observed between the CSF levels of HVA and 5-HIAA, and the concentrations of their precursor amino acids in either plasma or CSF. The MOPEG level in CSF correlated positively with the plasma concentrations of several amino acids.
Sigma-Aldrich
甘氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L -谷氨酰胺 溶液, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
甘氨酸, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-谷氨酰胺溶液 200mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
甘氨酸, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
甘氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L -赖氨酸, ≥98% (TLC)
Sigma-Aldrich
L-丝氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-异亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-谷氨酰胺, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Sigma-Aldrich
L-天冬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-天冬酰胺, ≥98% (HPLC)
Sigma-Aldrich
L -缬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-亮氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
甘氨酸, ACS reagent, ≥98.5%
Sigma-Aldrich
L-丙氨酸, ≥98% (TLC)
Sigma-Aldrich
L-异亮氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苏氨酸, reagent grade, ≥98% (HPLC)
SAFC
L-甲硫氨酸
SAFC
L-谷氨酰胺
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-丝氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
甘氨酸, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)