  • Search Results
  • Reduced cerebrospinal fluid 5-hydroxyindoleacetic acid and homovanillic acid in children with epilepsy.
Reduced cerebrospinal fluid 5-hydroxyindoleacetic acid and homovanillic acid in children with epilepsy.
Neurology (1975-01-01)
B A Shaywitz, D J Cohen, M B Bowers
PMID803305
ABSTRACT
Cerebrospinal fluid (CSF) concentrations of 5-hydroxyindoleacetic acid (5-HIAA), homovanillic acid (HVA) and probenecid were examined in 14 children with epilepsy (ages 6 months to 17 years) and 17 controls (ages 14 months to 16 years). The concentrations of amine metabolites were significantly correlated with probenecid concentrations in both groups of children. Cerebrospinal fluid concentrations of 5-HIAA were 63.6 ng per milliliter plus or minus 8.23 S.E.M. and 117 ng per milliliter plus or minus 11.6 S.E.M. for the epilepsy and control groups respectively. HVA averaged 89.1 ng per milliliter plus or minus 15.2 S.E.M in epilepsy and 172 ng per milliliter plus or minus 19.2 S.E.M. in the control group. These findings indicate a significant difference between epilepsy and control groups. Probenecid concentrations were similar in each group. The reduced cerebrospinal fluid amine metabolite concentrations in children with epilepsy were not related to age, anticonvulsant medication, cerebrospinal fluid folate or protein concentration, or cerebrospinal fluid cell count. Our findings suggest a relationship between brain amines and epilepsy.
Sigma-Aldrich
L -谷氨酰胺 溶液, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-谷氨酰胺, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-谷氨酸, ReagentPlus®, ≥99% (HPLC)
SAFC
L-谷氨酰胺溶液 200mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
L-异亮氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-异亮氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-亮氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-谷氨酰胺, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-甲硫氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-谷氨酰胺
Sigma-Aldrich
γ-氨基丁酸, ≥99%
SAFC
L-甲硫氨酸
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Sigma-Aldrich
L-谷氨酸, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-苏氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-天冬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-天冬酰胺, ≥98% (HPLC)
Sigma-Aldrich
L-苏氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-丙氨酸, ≥98% (TLC)
Sigma-Aldrich
L -缬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L -缬氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
腺苷 5′-二磷酸, ≥95% (HPLC)
Sigma-Aldrich
L-瓜氨酸, ≥98% (TLC)
SAFC
L-酪氨酸
SAFC
L-异亮氨酸