  • Search Results
  • Adding an unnatural covalent bond to proteins through proximity-enhanced bioreactivity.
Adding an unnatural covalent bond to proteins through proximity-enhanced bioreactivity.
Nature methods (2013-08-06)
Zheng Xiang, Haiyan Ren, Ying S Hu, Irene Coin, Jing Wei, Hu Cang, Lei Wang
ABSTRACT
Natural proteins often rely on the disulfide bond to covalently link side chains. Here we genetically introduce a new type of covalent bond into proteins by enabling an unnatural amino acid to react with a proximal cysteine. We demonstrate the utility of this bond for enabling irreversible binding between an affibody and its protein substrate, capturing peptide-protein interactions in mammalian cells, and improving the photon output of fluorescent proteins.
Sigma-Aldrich
单克隆抗-FLAG® M2-过氧化物酶(HRP) 小鼠抗, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-苯基丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-苯基丙氨酸, 99%
Supelco
L-苯基丙氨酸, certified reference material, TraceCERT®
Supelco
L-苯基丙氨酸, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC