  • Search Results
  • Allosteric inhibitor specificity of Thermotoga maritima 3-deoxy-D-arabino-heptulosonate 7-phosphate synthase.
Allosteric inhibitor specificity of Thermotoga maritima 3-deoxy-D-arabino-heptulosonate 7-phosphate synthase.
FEBS letters (2013-08-07)
Penelope J Cross, Emily J Parker
ABSTRACT
3-Deoxy-D-arabino-heptulosonate 7-phosphate synthase (DAH7PS) catalyses the first step of the shikimate pathway for the biosynthesis of aromatic amino acids. Allosteric regulation of Thermotoga maritima DAH7PS is mediated by L-Tyr binding to a discrete ACT regulatory domain appended to a core catalytic (β/α)8 barrel. Variants of T. maritima DAH7PS (TmaDAH7PS) were created to probe the role of key residues in inhibitor selection. Substitution Ser31Gly severely reduced inhibition by L-Tyr. In contrast both L-Tyr and L-Phe inhibited the TmaHis29Ala variant, while the variant where Ser31 and His29 were interchanged (His29Ser/Ser31His), was inhibited to a greater extent by L-Phe than L-Tyr. These studies highlight the role and importance of His29 and Ser31 for determining both inhibitory ligand selectivity and the potency of allosteric response by TmaDAH7PS.
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Supelco
L-苯基丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-苯基丙氨酸, 99%
Supelco
L-苯基丙氨酸, certified reference material, TraceCERT®
Supelco
L-苯基丙氨酸, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC
SAFC
L-苯基丙氨酸