Merck
  • 主页
  • 查找结果
  • A CRYGC gene mutation associated with autosomal dominant pulverulent cataract.
A CRYGC gene mutation associated with autosomal dominant pulverulent cataract.
Gene (2013-08-21)
Luz Ma González-Huerta, Olga Messina-Baas, Héctor Urueta, Jaime Toral-López, Sergio A Cuevas-Covarrubias
摘要
To describe at molecular level a family with pulverulent congenital cataract associated with a CRYGC gene mutation. One family with several affected members with pulverulent congenital cataract and 230 healthy controls were examined. Genomic DNA from leukocytes was isolated to analyze the CRYGA-D cluster, CX46, CX50 and MIP genes through high-resolution melting curve and DNA sequencing. DNA sequencing in the affected members revealed the c.143G>A mutation (p.R48H) in exon 2 of the CRYGC gene; 230 healthy controls and ten healthy relatives were also analyzed and none of them showed the c.143G>A mutation. No other polymorphisms or mutations were found to be present. In the present study, we described a family with pulverulent congenital cataract that segregated the c.143G>A mutation (p.R48H) in the CRYGC gene. A few mutations have been described in the CRYGC gene in autosomal dominant cataract, none of them with pulverulent cataract making clear the clinical heterogeneity of congenital cataract. This mutation has been associated with the phenotype of congenital cataract but also is considered an SNP in the NCBI data base. Our data and previous report suggest that p.R48H could be a disease-causing mutation and not an SNP.
材料
货号
品牌
产品描述
Sigma-Aldrich
L-组氨酸, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-组氨酸, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
SAFC
L-组氨酸
Sigma-Aldrich
L-组氨酸, BioUltra, ≥99.5% (NT)
Supelco
L-组氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-组氨酸, certified reference material, TraceCERT®