Merck
A missense mutation in DUSP6 is associated with Class III malocclusion.
Journal of dental research (2013-08-24)
T Nikopensius, M Saag, T Jagomägi, T Annilo, M Kals, P A Kivistik, L Milani, A Metspalu
ABSTRACT
Class III malocclusion is a common dentofacial phenotype with a variable prevalence according to ethnic background. The etiology of Class III malocclusion has been attributed mainly to interactions between susceptibility genes and environmental factors during the morphogenesis of the mandible and maxilla. Class III malocclusion shows familial recurrence, and family-based studies support a predominance of an autosomal-dominant mode of inheritance. We performed whole-exome sequencing on five siblings from an Estonian family affected by Class III malocclusion. We identified a rare heterozygous missense mutation, c.545C>T (p.Ser182Phe), in the DUSP6 gene, a likely causal variant. This variant co-segregated with the disease following an autosomal-dominant mode of inheritance with incomplete penetrance. Transcriptional activation of DUSP6 has been presumed to be regulated by FGF/FGFR and MAPK/ERK signaling during fundamental processes at early stages of skeletal development. Several candidate genes within a linkage region on chromosome 12q22-q23--harboring DUSP6--are implicated in the regulation of maxillary or mandibular growth. The current study reinforces that the 12q22-q23 region is biologically relevant to craniofacial development and may be genetically linked to the Class III malocclusion.
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Supelco
L-苯基丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-苯基丙氨酸, 99%
Supelco
L-苯基丙氨酸, certified reference material, TraceCERT®
Supelco
L-苯基丙氨酸, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC
SAFC
L-苯基丙氨酸