Merck
  • 主页
  • 查找结果
  • qPCR increases sensitivity to detect cytomegalovirus in formalin-fixed, paraffin-embedded tissue of gastrointestinal biopsies.
qPCR increases sensitivity to detect cytomegalovirus in formalin-fixed, paraffin-embedded tissue of gastrointestinal biopsies.
Human pathology (2013-10-22)
Morgan H McCoy, Kristin Post, Joyashree D Sen, Hsim Y Chang, Zijin Zhao, Rong Fan, Shaoxiong Chen, Diane Leland, Liang Cheng, Jingmei Lin
摘要
Histopathologic diagnosis of gastrointestinal (GI) tract cytomegaloviral (CMV) infection relies on hematoxylin and eosin (H&E)-stained tissue, along with the aid of immunohistochemistry (IHC). However, non-classic appearing inclusions or atypical IHC staining patterns remain an ongoing concern for pathologists. We reported the use of real-time polymerase chain reaction (qPCR) on nucleic acid extracted from paraffin-embedded, formalin-fixed tissue of GI biopsies from cases of CMV infection (n = 91) diagnosed by H&E and IHC. Seventy-nine biopsies, including normal colon biopsies (n = 35), active colitis (n = 25), and active duodenitis (n = 19), were used as negative controls. Of 91 CMV-positive biopsies diagnosed by histology, 88 tested positive by qPCR, with a sensitivity of 96.7%. Of 79 negative controls, 78 were negative and 1 positive by qPCR, resulting in a specificity of 98.7%. Of the cases that were positive for CMV by histopathology, there were an additional 40 biopsies taken from these patients either during the same or previous procedures, some taken just days prior, which were negative for CMV by histology. Interestingly, 22 (55%) of these biopsies tested positive by qPCR, which correlated well with additional clinical CMV results. By defining qPCR as the "gold standard" for a CMV result, histology (H&E and/or IHC) had a sensitivity and specificity of 79% and 97%, respectively. Eighteen biopsies were found negative by H&E and equivocal by IHC. Among them, 14 (78%) tested positive for CMV by qPCR, which also correlated well with additional clinical results. qPCR is a sensitive, specific, and rapid molecular tool that may be helpful to aid in early diagnosis of CMV infection on equivocal or clinically highly suspicious small GI biopsies.
材料
货号
品牌
产品描述
Sigma-Aldrich
甲醛 溶液, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
甲醛 溶液, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
甲醛 溶液, for molecular biology, BioReagent, ≥36.0% in H2O (T)
SAFC
甲醛 溶液, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
甲醛 溶液, meets analytical specification of USP, ≥34.5 wt. %
Supelco
甲醛 溶液, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
甲醛 溶液, tested according to Ph. Eur.