  • Search Results
  • Perfluorooctanesulfonate (PFOS) perturbs male rat Sertoli cell blood-testis barrier function by affecting F-actin organization via p-FAK-Tyr(407): an in vitro study.
Perfluorooctanesulfonate (PFOS) perturbs male rat Sertoli cell blood-testis barrier function by affecting F-actin organization via p-FAK-Tyr(407): an in vitro study.
Endocrinology (2013-10-31)
Hin-Ting Wan, Dolores D Mruk, Chris K C Wong, C Yan Cheng
ABSTRACT
Environmental toxicants such as perfluorooctanesulfonate (PFOS) have been implicated in male reproductive dysfunction, including reduced sperm count and semen quality, in humans. However, the underlying mechanism(s) remains unknown. Herein PFOS at 10-20 μM (∼5-10 μg/mL) was found to be more potent than bisphenol A (100 μM) in perturbing the blood-testis barrier (BTB) function by disrupting the Sertoli cell tight junction-permeability barrier without detectable cytotoxicity. We also delineated the underlying molecular mechanism by which PFOS perturbed Sertoli cell BTB function using an in vitro model that mimics the BTB in vivo. First, PFOS perturbed F-actin organization in Sertoli cells, causing truncation of actin filaments at the BTB. Thus, the actin-based cytoskeleton was no longer capable of supporting the distribution and/or localization of actin-regulatory and adhesion proteins at the cell-cell interface necessary to maintain BTB integrity. Second, PFOS was found to perturb inter-Sertoli cell gap junction (GJ) communication based on a dye-transfer assay by down-regulating the expression of connexin-43, a GJ integral membrane protein. Third, phosphorylated focal adhesion kinase (FAK)-Tyr(407) was found to protect the BTB from the destructive effects of PFOS as shown in a study via an overexpression of an FAK Y407E phosphomimetic mutant. Also, transfection of Sertoli cells with an FAK-specific microRNA, miR-135b, to knock down the expression of phosphorylated FAK-Tyr(407) was found to worsen PFOS-mediated Sertoli cell tight junction disruption. In summary, PFOS-induced BTB disruption is mediated by down-regulating phosphorylated FAK-Tyr(407) and connexin-43, which in turn perturbed F-actin organization and GJ-based intercellular communication, leading to mislocalization of actin-regulatory and adhesion proteins at the BTB.
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
Supelco
双酚 A, ≥99%
SAFC
L-酪氨酸
Sigma-Aldrich
L-酪氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
十七氟辛烷磺酸 溶液, ~40% in H2O (T)
Sigma-Aldrich
L-酪氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
双酚 A, 97%
Supelco
十七氟辛烷磺酸 溶液, 100 μg/mL in methanol, analytical standard
Supelco
L-酪氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-酪氨酸, FG
Sigma-Aldrich
抗ARP3抗体,小鼠单克隆 小鼠抗, clone FMS338, purified from hybridoma cell culture
Supelco
L-酪氨酸, certified reference material, TraceCERT®