• 主页
  • 查找结果
  • Effects of copper and aluminum on the adsorption of sulfathiazole and tylosin on peat and soil.
Effects of copper and aluminum on the adsorption of sulfathiazole and tylosin on peat and soil.
Environmental pollution (Barking, Essex : 1987) (2013-11-10)
Zhiguo Pei, Shuang Yang, Lingyun Li, Chunmei Li, Shuzhen Zhang, Xiao-quan Shan, Bei Wen, Baoyuan Guo
摘要
Effects of copper (Cu) and aluminum (Al) on the adsorption of sulfathiazole (STZ) and tylosin (T) to peat and soil were investigated using a batch equilibration method. Results show that Cu suppressed STZ adsorption onto peat and soil at pH < 5.0 because of the electrostatic competition, while increased STZ adsorption at pH > 5.0 due to the formation of STZ-Cu complexes and/or Cu bridge. In contrast, Al only decreased STZ adsorption at pH < 6.0, and exerted slight effect on STZ adsorption at >6.0. As for T, both Cu and Al suppressed its adsorption over the entire pH range owing to three reasons: 1) electrostatic competition between Cu/Al and T(+); 2) Cu/Al adsorption made the soil and peat surface less negatively charged, which was unfavorable for T(+) adsorption; 3) the shrunken pore size of peat and soil retarded the diffusion of large-sized T into these pores.
材料
货号
品牌
产品描述
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铝, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
铝, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Supelco
磺胺噻唑, VETRANAL®, analytical standard
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Supelco
磺胺噻唑
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铝, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铝, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铝, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
铝, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
磺胺噻唑 钠盐, ≥99%
Sigma-Aldrich
铝, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
铝, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Sigma-Aldrich
铝, pellets, 3-12 mm, 99.99% trace metals basis