  • Search Results
  • Myofibrillar muscle protein synthesis rates subsequent to a meal in response to increasing doses of whey protein at rest and after resistance exercise.
Myofibrillar muscle protein synthesis rates subsequent to a meal in response to increasing doses of whey protein at rest and after resistance exercise.
The American journal of clinical nutrition (2013-11-22)
Oliver C Witard, Sarah R Jackman, Leigh Breen, Kenneth Smith, Anna Selby, Kevin D Tipton
ABSTRACT
The intake of whey, compared with casein and soy protein intakes, stimulates a greater acute response of muscle protein synthesis (MPS) to protein ingestion in rested and exercised muscle. We characterized the dose-response relation of postabsorptive rates of myofibrillar MPS to increasing amounts of whey protein at rest and after exercise in resistance-trained, young men. Volunteers (n = 48) consumed a standardized, high-protein (0.54 g/kg body mass) breakfast. Three hours later, a bout of unilateral exercise (8 × 10 leg presses and leg extensions; 80% one-repetition maximum) was performed. Volunteers ingested 0, 10, 20, or 40 g whey protein isolate immediately (~10 min) after exercise. Postabsorptive rates of myofibrillar MPS and whole-body rates of phenylalanine oxidation and urea production were measured over a 4-h postdrink period by continuous tracer infusion of labeled [(13)C6] phenylalanine and [(15)N2] urea. Myofibrillar MPS (mean ± SD) increased (P < 0.05) above 0 g whey protein (0.041 ± 0.015%/h) by 49% and 56% with the ingestion of 20 and 40 g whey protein, respectively, whereas no additional stimulation was observed with 10 g whey protein (P > 0.05). Rates of phenylalanine oxidation and urea production increased with the ingestion of 40 g whey protein. A 20-g dose of whey protein is sufficient for the maximal stimulation of postabsorptive rates of myofibrillar MPS in rested and exercised muscle of ~80-kg resistance-trained, young men. A dose of whey protein >20 g stimulates amino acid oxidation and ureagenesis. This trial was registered at http://www.isrctn.org/ as ISRCTN92528122.
Sigma-Aldrich
L-苯基丙氨酸, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-苯基丙氨酸, reagent grade, ≥98%
Supelco
L-苯基丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-苯基丙氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-苯基丙氨酸, 99%
Supelco
L-苯基丙氨酸, certified reference material, TraceCERT®
Supelco
L-苯基丙氨酸, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Sigma-Aldrich
L-苯基丙氨酸, 99%, FCC
SAFC
L-苯基丙氨酸