Merck
  • 主页
  • 查找结果
  • Nutrient accumulation in various plant parts of dominant tree species of three different localities.
Nutrient accumulation in various plant parts of dominant tree species of three different localities.
Pakistan journal of biological sciences : PJBS (2014-02-08)
Beena Sharma, Kavita Sharma
摘要
In the present study, accumulation of nutrients (N, P, Ca, Mg, and K) in various plant parts of three different dominant trees i.e., Acacia senegal (As), Acacia tortilis (At) and Eucalyptus camaldulensis (Ec) was estimated. Concentration of nutrients was estimated for bole, first order branch, other branch, foliage, seeds, stump root, lateral root and fine roots. The study was carried out in the sand dunes of Western foot hill gaps of Central Aravalli located at a distance of 10 km n-w to Ajmer, a centrally situated city of Rajasthan. The concentration of nutrients was found to be maximum in foliage except for nitrogen which was estimated higher in seeds of A. senegal and A. tortilis. A different pattern was recorded for E. camaldulensis where N, Ca, and Mg were recorded maximum in other branch, while p and K in foliage parts. Result shows the concentration of nutrients in different tree components in the order: foliage>seeds> their branch>first order branch>bole, and in root components: fine root>lateral root>stump root. Total nutrient concentration was found to be maximum in As followed by At and Ec. It is concluded that the foliage component of various trees has maximum nutrient concentrations. There are marked variations in the concentration of different nutrients in each component.
材料
货号
品牌
产品描述
Sigma-Aldrich
镁, turnings, reagent grade, 98%
Sigma-Aldrich
红磷, ≥97.0%
Sigma-Aldrich
红磷, ≥99.99% trace metals basis
Sigma-Aldrich
镁, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
镁, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
镁, grit, ≥99.0% (KT)
Sigma-Aldrich
镁, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
镁, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol