  • Search Results
  • Polymer-Supported Hantzsch 1, 4-Dihydropyridine Ester: An Efficient Biomimetic Hydrogen Source.
Polymer-Supported Hantzsch 1, 4-Dihydropyridine Ester: An Efficient Biomimetic Hydrogen Source.
Advanced Synthesis & Catalysis (2008)
He R, et al.