  • Search Results
  • Inhibiting tyrosine phosphorylation of protein kinase Cδ (PKCδ) protects the salivary gland from radiation damage.
Inhibiting tyrosine phosphorylation of protein kinase Cδ (PKCδ) protects the salivary gland from radiation damage.
The Journal of biological chemistry (2014-02-27)
Sten M Wie, Tariq S Adwan, James DeGregori, Steven M Anderson, Mary E Reyland
ABSTRACT
Radiation therapy for head and neck cancer can result in extensive damage to normal adjacent tissues such as the salivary gland and oral mucosa. We have shown previously that tyrosine phosphorylation at Tyr-64 and Tyr-155 activates PKCδ in response to apoptotic stimuli by facilitating its nuclear import. Here we have identified the tyrosine kinases that mediate activation of PKCδ in apoptotic cells and have explored the use of tyrosine kinase inhibitors for suppression of irradiation-induced apoptosis. We identify the damage-inducible kinase, c-Abl, as the PKCδ Tyr-155 kinase and c-Src as the Tyr-64 kinase. Depletion of c-Abl or c-Src with shRNA decreased irradiation- and etoposide-induced apoptosis, suggesting that inhibitors of these kinases may be useful therapeutically. Pretreatment with dasatinib, a broad spectrum tyrosine kinase inhibitor, blocked phosphorylation of PKCδ at both Tyr-64 and Tyr-155. Expression of "gate-keeper" mutants of c-Abl or c-Src that are active in the presence of dasatinib restored phosphorylation of PKCδ at Tyr-155 and Tyr-64, respectively. Imatinib, a c-Abl-selective inhibitor, also specifically blocked PKCδ Tyr-155 phosphorylation. Dasatinib and imatinib both blocked binding of PKCδ to importin-α and nuclear import, demonstrating that tyrosine kinase inhibitors can inhibit nuclear accumulation of PKCδ. Likewise, pretreatment with dasatinib also suppressed etoposide and radiation induced apoptosis in vitro. In vivo, pre-treatment of mice with dasatinib blocked radiation-induced apoptosis in the salivary gland by >60%. These data suggest that tyrosine kinase inhibitors may be useful prophylactically for protection of nontumor tissues in patients undergoing radiotherapy of the head and neck.
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
SAFC
L-酪氨酸
Sigma-Aldrich
L-酪氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-酪氨酸, BioUltra, ≥99.0% (NT)
Supelco
L-酪氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
酪氨酸, European Pharmacopoeia (EP) Reference Standard
USP
L-酪氨酸, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-酪氨酸, FG
Millipore
Protein A–Sepharose® 6MB, aqueous ethanol suspension
Supelco
L-酪氨酸, certified reference material, TraceCERT®