Merck
Catalytic upgrading of xylan over mesoporous Y catalyst.
Journal of nanoscience and nanotechnology (2014-04-17)
Seong-Soo Kim, Bo Ram Jun, Sung Hoon Park, Jong-Ki Jeon, Dong Jin Suh, Tae-Wan Kim, Young-Kwon Park
ABSTRACT
In-situ catalytic cracking of xylan, a model compound of hemicellulose, was carried out using pyrolysis-gas chromatography/mass spectrometry over mesoporous Y for the first time. Experiments were conducted at three different temperatures, 400 degrees C, 450 degrees C, and 500 degrees C, to investigate the effect of reaction temperature. Three different biomass-to-catalyst ratios, 1:1, 1:2, and 1:3, were tested at 500 degrees C to examine the effect of catalyst dose. In addition, the catalytic activity of mesoporous Y was compared with that of Al-MCM-41. The catalysts used were characterized by N2 adsorption-desorption, temperature programmed desorption of NH3, and X-ray diffraction. The main pyrolysis products of xylan were acids, hydrocarbons, phenolics, oxygenates, aromatics, and polycyclic aromatic hydrocarbons. Mesoporous Y, which has acid sites with larger quantity and stronger acidity than those of Al-MCM-41, was shown to enhance the quality of bio-oil to a larger extent, producing a larger quantity of high-value-added products, such as aromatics and furans.
Sigma-Aldrich
二氧化硅, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
沸石
Sigma-Aldrich
LUDOX® HS-40胶态氧化硅, 40 wt. % suspension in H2O
Sigma-Aldrich
二氧化硅, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
LUDOX ® AS-40 胶态二氧化硅, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 胶态氧化硅, 50 wt. % suspension in H2O
Sigma-Aldrich
二氧化硅, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
二氧化硅, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
Sigma-Aldrich
介孔结构的二氧化硅, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX ® AS-30 胶态二氧化硅, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX®SM胶体二氧化硅, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX ® AM 胶态二氧化硅, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX ® TMA 胶态二氧化硅, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX ® HS-30 胶态二氧化硅, 30 wt. % suspension in H2O
Sigma-Aldrich
介孔结构的硅铝酸盐, MCM-41 (hexagonal)
Sigma-Aldrich
LUDOX ® TM-40 胶态二氧化硅, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL 胶态氧化硅, 30 wt. % suspension in H2O
Sigma-Aldrich
二氧化硅
Sigma-Aldrich
LUDOX® LS 胶态氧化硅, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X 胶态氧化硅, 45 wt. % suspension in H2O
Sigma-Aldrich
介孔结构的二氧化硅, MSU-F (cellular foam)
Sigma-Aldrich
二氧化硅, MSU-H (large pore 2D hexagonal)
Sigma-Aldrich
介孔结构的二氧化硅, HMS (wormhole)