  • Search Results
  • Removal of copper ions from water using epichlorohydrin cross-linked beta-cyclodextrin polymer: characterization, isotherms and kinetics.
Removal of copper ions from water using epichlorohydrin cross-linked beta-cyclodextrin polymer: characterization, isotherms and kinetics.
Water environment research : a research publication of the Water Environment Federation (2014-05-24)
M Tajuddin Sikder, Md Shariful Islam, Tohru Kikuchi, Junichi Suzuki, Takeshi Saito, Masaaki Kurasaki
ABSTRACT
Beta-cyclodextrin (beta-CD) cross-linked with epichlorohydrin to form water insoluble beta-cyclodextrin polymer (beta-CDP) has been shown to be an effective sorbent for sorption of organic particles, but the sorption of copper (Cu2+) in aqueous solutions by beta-CDP has not been conducted. The objective of this study was to explore the sorption mechanism of beta-CDP for copper. The effects of different experimental conditions such as pH, ionic strength, contact time, and temperature were inspected using a batch method. In addition, binding scheme was estimated by using Fourier transform infrared (FTIR) spectroscopy, Xray photoelectron spectroscopy (XPS), a scanning electron microscope (SEM), and Brunauer-Emmett-Teller (BET) analysis. The adsorption of Cu2+ was observed to be higher at pH 6.0. The kinetic study revealed that the adsorption is fitted well by the pseudo-second-order kinetic model. The maximum binding of Cu2+ was estimated to be 111.11 mg/g through the Langmuir isotherm model--much higher than the existing sorption technologies. Hence, the adsorption-desorption trends of epichlorohydrin cross-linked with beta-CD, along with its good recyclability, establish an alternative, effective, and novel remediation technology for the removal of Cu2+ from aqueous solutions.
Sigma-Aldrich
β-环糊精, ≥97%
Sigma-Aldrich
阿特拉津 (莠去津), purum, ≥99% (GC)
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Supelco
阿特拉津 (莠去津), analytical standard
Sigma-Aldrich
(S)-(+)-环氧氯丙烷, 98%
Sigma-Aldrich
β-环糊精, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
β-环糊精
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
USP
β-环糊精, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Supelco
环糊精, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-环糊精, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, ≥98.0% (HPLC)
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
β-环糊精, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
β-环糊精, produced by Wacker Chemie AG, Burghausen, Germany, ≥95.0% (HPLC)
Sigma-Aldrich
铜, wire, diam. 0.64 mm, 99.995% trace metals basis