Merck
Occurrence of a taurine derivative in an antarctic glass sponge.
Natural product communications (2014-05-30)
Marianna Carbone, Laura Núñez-Pons, M Letizia Ciavatta, Francesco Castelluccio, Conxita Avila, Margherita Gavagnin
ABSTRACT
The n-butanol extract of an Antarctic hexactinellid sponge, Anoxycalyx (Scolymastra) joubini, was found to contain a taurine-conjugated anthranilic acid, never reported so far either as a natural product or by synthesis. The compound was inactive against human cancer cells in an in vitro growth inhibitory test, and also showed no antibacterial activity.
Sigma-Aldrich
牛磺酸, ≥99%
Sigma-Aldrich
牛磺酸, suitable for cell culture, meets USP testing specifications
Supelco
牛磺酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
牛磺酸, BioUltra, ≥99.5% (T)
SAFC
牛磺酸
Sigma-Aldrich
牛磺酸, ≥98%, FG