Merck
The link between copper and Wilson's disease.
Science progress (2013-11-20)
Rupert Purchase
ABSTRACT
Wilson's disease (hepatolenticular degeneration) is a rare inherited autosomal recessive disorder of copper metabolism leading to copper accumulation in the liver and extrahepatic organs such as the brain and cornea. Patients may present with combinations of hepatic, neurological and psychiatric symptoms. Copper is the therapeutic target for the treatment of Wilson's disease. But how did copper come to be linked with Wilson's disease? The answer encompasses a study of enzootic neonatal ataxia in lambs in the 1930s, the copper-chelating properties of British Anti-Lewisite, and the chemical analysis for copper of the organs of deceased Wilson's disease patients in the mid-to-late 1940s. Wilson's disease is one of a number of copper-related disorders where loss of copper homeostasis as a result of genetic, nutritional or environmental factors affects human health.
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
铜, electrolytic, Analytical Reagent, reag. Ph. Eur., ≥99.9% (complexometric), wire
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
铜, shot, −3-+14 mesh, 99%
Sigma-Aldrich
铜, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
铜, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%
Sigma-Aldrich
铜, wire, diam. 2.0 mm, 99.999% trace metals basis
铜, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
铜, mesh, 130x130mm, nominal aperture 0.2mm, thickness 0.004mm, wire diameter 0.0125mm, 118 wires/inch, open area 88%, electro-formed mesh
铜, foil, 100x100mm, thickness 0.125mm, hard, 99.95+%