• 主页
  • 查找结果
  • Advances in fractal germanium micro/nanoclusters induced by gold: microstructures and properties.
Advances in fractal germanium micro/nanoclusters induced by gold: microstructures and properties.
Journal of nanoscience and nanotechnology (2014-04-23)
Zhiwen Chen, Chan-Hung Shek, C M Lawrence Wu, Joseph K L Lai
摘要
Germanium materials are a class of unique semiconductor materials with widespread technological applications because of their valuable semiconducting, electrical, optical, and thermoelectric power properties in the fields of macro/mesoscopic materials and micro/nanodevices. In this review, we describe the efforts toward understanding the microstructures and various properties of the fractal germanium micro/nanoclusters induced by gold prepared by high vacuum thermal evaporation techniques, highlighting contributions from our laboratory. First, we present the integer and non-integer dimensional germanium micro/nanoclusters such as nanoparticles, nanorings, and nanofractals induced by gold and annealing. In particular, the nonlinear electrical behavior of a gold/germanium bilayer film with the interesting nanofractal is discussed in detail. In addition, the third-order optical nonlinearities of the fractal germanium nanocrystals embedded in gold matrix will be summarized by using the sensitive and reliable Z-scan techniques aimed to determine the nonlinear absorption coefficient and nonlinear refractive index. Finally, we emphasize the thermoelectric power properties of the gold/germanium bilayer films. The thermoelectric power measurement is considered to be a more effective method than the conductivity for investigating superlocalization in a percolating system. This research may provide a novel insight to modulate their competent performance and promote rational design of micro/nanodevices. Once mastered, germanium thin films with a variety of fascinating micro/nanoclusters will offer vast and unforeseen opportunities in the semiconductor industry as well as in other fields of science and technology.
材料
货号
品牌
产品描述
Sigma-Aldrich
金, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
金, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
金, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
镀金显微镜载片, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
金, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
金, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
金, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
镀金显微镜载片, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
金, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
金, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
金, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
金, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
金, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
金, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
金, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
金, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
锗, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
锗, chips, 99.999% trace metals basis
Sigma-Aldrich
金, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
金, foil, thickness 0.25 mm, ≥99.9% trace metals basis
金, sphere, 50pcs, diameter 0.5mm, 99.99%
金, sphere, 200pcs, diameter 0.5mm, 99.99%
Sigma-Aldrich
金, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
金, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
金, foil, thickness 0.05 mm, 99.99% trace metals basis
锗, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
Sigma-Aldrich
金, wire, diam. 0.127 mm, 99.99% trace metals basis
锗, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
金, insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%