• 主页
  • 查找结果
  • HIF-1α-induced HSP70 regulates anabolic responses in articular chondrocytes under hypoxic conditions.
HIF-1α-induced HSP70 regulates anabolic responses in articular chondrocytes under hypoxic conditions.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society (2014-03-29)
Shinji Tsuchida, Yuji Arai, Kenji A Takahashi, Tsunao Kishida, Ryu Terauchi, Kuniaki Honjo, Shuji Nakagawa, Hiroaki Inoue, Kazuya Ikoma, Keiichiro Ueshima, Tomohiro Matsuki, Osam Mazda, Toshikazu Kubo
摘要
We assessed whether heat shock protein 70 (HSP70) is involved in hypoxia inducible factor 1 alpha (HIF-1α)-dependent anabolic pathways in articular chondrocytes under hypoxic conditions. Primary rabbit chondrocytes were cultured under normoxia (20% oxygen condition) or hypoxia (1% oxygen condition). Alternatively, cells cultured under normoxia were treated with CoCl2 , which induces HIF-1α, to simulate hypoxia, or transfected with siRNAs targeting HIF-1α (si-HIF-1α) and HSP70 (si-HSP70) under hypoxia. HSP70 expression was enhanced by the increased expression of HIF-1α under hypoxia or simulated hypoxia, but not in the presence of si-HIF-1α. Hypoxia-induced overexpression of ECM genes was significantly suppressed by si-HIF-1α or si-HSP70. Cell viability positively correlated with hypoxia, but transfection with si-HIF-1α or si-HSP70 abrogated the chondroprotective effects of hypoxia. Although LDH release from sodium nitroprusside-treated cells and the proportion of TUNEL positive cells were decreased under hypoxia, transfection with si-HIF-1α or si-HSP70 almost completely blocked these effects. These findings indicated that HIF-1α-induced HSP70 overexpression increased the expression levels of ECM genes and cell viability, and protected chondrocytes from apoptosis. HIF-1α may regulate the anabolic effects of chondrocytes under hypoxic conditions by regulating HSP70 expression.
材料
货号
品牌
产品描述
Sigma-Aldrich
二氯化钴, 97%
Sigma-Aldrich
0.1 M 氯化钴 溶液
Sigma-Aldrich
二氯化钴, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
钴, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
钴, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
二氯化钴, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
二氯化钴, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
钴, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
钴, granular, 99.99% trace metals basis
Sigma-Aldrich
钴, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
钴, pieces, 99.5% trace metals basis
Sigma-Aldrich
钴, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
钴, foil, thickness 0.25 mm, ≥99.99% trace metals basis
钴, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%
钴, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
钴, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
钴, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
钴, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
钴, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
钴, foil, 4mm disks, thickness 0.003mm, 99.9%
钴, foil, 4mm disks, thickness 0.004mm, 99.9%
钴, foil, 4mm disks, thickness 0.005mm, 99.9%
钴, foil, 4mm disks, thickness 0.006mm, 99.9%
钴, foil, 4mm disks, thickness 0.007mm, 99.9%
钴, foil, 4mm disks, thickness 0.008mm, 99.9%
钴, foil, 4mm disks, thickness 0.009mm, 99.9%
钴, foil, 4mm disks, thickness 0.0125mm, 99.9%
钴, foil, 4mm disks, thickness 0.015mm, 99.9%
钴, foil, 4mm disks, thickness 0.01mm, 99.9%
钴, foil, 4mm disks, thickness 0.025mm, 99.9%