Merck
  • 主页
  • 查找结果
  • Low resolution X-ray structure of γ-glutamyltranspeptidase from Bacillus licheniformis: opened active site cleft and a cluster of acid residues potentially involved in the recognition of a metal ion.
Low resolution X-ray structure of γ-glutamyltranspeptidase from Bacillus licheniformis: opened active site cleft and a cluster of acid residues potentially involved in the recognition of a metal ion.
Biochimica et biophysica acta (2014-05-02)
Long-Liu Lin, Yi-Yu Chen, Meng-Chun Chi, Antonello Merlino
摘要
γ-Glutamyltranspeptidases (γ-GTs) cleave the γ-glutamyl amide bond of glutathione and transfer the released γ-glutamyl group to water (hydrolysis) or acceptor amino acids (transpeptidation). These ubiquitous enzymes play a key role in the biosynthesis and degradation of glutathione, and in xenobiotic detoxification. Here we report the 3Å resolution crystal structure of Bacillus licheniformis γ-GT (BlGT) and that of its complex with l-Glu. X-ray structures confirm that BlGT belongs to the N-terminal nucleophilic hydrolase superfamily and reveal that the protein possesses an opened active site cleft similar to that reported for the homologous enzyme from Bacillus subtilis, but different from those observed for human γ-GT and for γ-GTs from other microorganisms. Data suggest that the binding of l-Glu induces a reordering of the C-terminal tail of BlGT large subunit and allow the identification of a cluster of acid residues that are potentially involved in the recognition of a metal ion. The role of these residues on the conformational stability of BlGT has been studied by characterizing the autoprocessing, enzymatic activity, chemical and thermal denaturation of four new Ala single mutants. The results show that replacement of Asp568 with an Ala affects both the autoprocessing and structural stability of the protein.
材料
货号
品牌
产品描述
Sigma-Aldrich
L-谷氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-谷氨酸, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-天冬氨酸, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-丙氨酸, ≥98% (TLC)
Sigma-Aldrich
镁, turnings, reagent grade, 98%
Sigma-Aldrich
镁, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
L-丙氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-谷氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-天冬氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-丙氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-天冬氨酸, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D -谷氨酸, ≥99% (TLC)
Supelco
L-谷氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-天冬氨酸, BioUltra, ≥99.5% (T)
谷氨酸, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL -天冬氨酸, ≥99% (TLC)
Sigma-Aldrich
L-丙氨酸, ≥99%
Sigma-Aldrich
L -天冬氨酸 钾盐, ≥98% (HPLC)
Supelco
L-丙氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-丙氨酸, ≥99.0% (NT)
SAFC
L-天冬氨酸
Sigma-Aldrich
γ-谷氨酰转肽酶 来源于马肾脏, Type VI, 5-12 units/mg solid
Sigma-Aldrich
L-谷氨酸 盐酸盐, ≥99% (HPLC)
Sigma-Aldrich
L-谷氨酸, FCC
天冬氨酸, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
镁, 20-230 mesh, reagent grade, 98%
丙氨酸, European Pharmacopoeia (EP) Reference Standard
Supelco
L-天冬氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-谷氨酸, certified reference material, TraceCERT®
Sigma-Aldrich
γ-Glutamyltranspeptidase from porcine kidney, Type IV, 2.0-6.0 units/mg solid