• 主页
  • 查找结果
  • Gene expression profiles reveal distinct immunological responses of cobalt and cerium dioxide nanoparticles in two in vitro lung epithelial cell models.
Gene expression profiles reveal distinct immunological responses of cobalt and cerium dioxide nanoparticles in two in vitro lung epithelial cell models.
Toxicology letters (2014-05-14)
Sandra Verstraelen, Sylvie Remy, Eudald Casals, Patrick De Boever, Hilda Witters, Antonietta Gatti, Victor Puntes, Inge Nelissen
摘要
Fragmentary knowledge exists on cellular signaling responses underlying possible adverse health effects of CoO- and CeO2-nanoparticles (NP)s after inhalation. We aimed to perform a time kinetic study of gene expression profiles induced by these NPs in alveolar A549 and bronchial BEAS-2B epithelial cells, and investigated possible immune system modulation. The kinetics of the cell responses induced by the NPs were different between the lung epithelial models. Both CoO- and CeO2-NP exposure induced mainly downregulation of gene transcription. BEAS-2B cells were found to be more sensitive, as they showed a higher number of differentially expressed transcripts (DET) at a 10-fold lower NP-concentration than A549 cells. Hierarchical clustering of all DET indicated that the transcriptional responses were heterogeneous among the two cell types and two NPs. Between 1% and 14% DET encoding markers involved in immune processes were observed. The transcriptional impact of the metal oxide NPs appeared to be cell-dependent, both at the general and immune response level, whereas each lung epithelial cell model responded differently to the two NP types. Thus, the study provides gene expression markers and immune processes involved in CoO- and CeO2-NP-induced toxicity, and demonstrates the usefulness of comprehensive-omics studies to differentiate between NP responses.
材料
货号
品牌
产品描述
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, ≥99.5% (titration)
Sigma-Aldrich
2-巯基乙醇, ≥99.0%
Sigma-Aldrich
2-巯基乙醇, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
氢化可的松, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES缓冲溶液, 1 M in H2O
Sigma-Aldrich
2-巯基乙醇, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
氢化可的松, ≥98% (HPLC)
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
氢化可的松, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
氧化铈 (IV) 氧化物, nanopowder, <25 nm particle size (BET)
Supelco
氢化可的松, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
氧化铈 (IV) 氧化物, powder, <5 μm, 99.9% trace metals basis
SAFC
羟乙基哌嗪乙硫磺酸
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
氧化铈 (IV) 氧化物, ≥99.0%
Supelco
2-巯基乙醇, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
氧化铈 (IV) 氧化物, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
钴, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
钴, powder, 2 μm particle size, 99.8% trace metals basis
氢化可的松, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
氧化铈 (IV) 氧化物, powder, 99.995% trace metals basis
Sigma-Aldrich
氧化铈(IV), nanoparticles, <25 nm particle size, 10 wt. % in H2O
Sigma-Aldrich
羟乙基哌嗪乙硫磺酸, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
氢化可的松, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
氢化可的松, meets USP testing specifications
峰鉴别用氢化可的松, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
铈, powder, −40 mesh, 99.9% trace rare earth metals basis