• 主页
  • 查找结果
  • Kinetic investigation of the rate-limiting step of manganese- and iron-lipoxygenases.
Kinetic investigation of the rate-limiting step of manganese- and iron-lipoxygenases.
Archives of biochemistry and biophysics (2014-05-27)
Anneli Wennman, Saeid Karkehabadi, Ernst H Oliw
摘要
Lipoxygenases (LOX) oxidize polyunsaturated fatty acids to hydroperoxides, which are generated by proton coupled electron transfer to the metal center with FeIIIOH- or MnIIIOH-. Hydrogen abstraction by FeIIIOH- of soybean LOX-1 (sLOX-1) is associated with a large deuterium kinetic isotope effect (D-KIE). Our goal was to compare the D-KIE and other kinetic parameters at different temperatures of sLOX-1 with 13R-LOX with catalytic manganese (13R-MnLOX). The reaction rate and the D-KIE of sLOX-1 with unlabeled and [11-2H2]18:2n-6 were almost temperature independent with an apparent D-KIE of ∼56 at 30°C, which is in agreement with previous studies. In contrast, the reaction rate of 13R-MnLOX increased 7-fold with temperature (8-50°C), and the apparent D-KIE decreased linearly from ∼38 at 8°C to ∼20 at 50°C. The kinetic lag phase of 13R-MnLOX was consistently extended at low temperatures. The Phe337Ile mutant of 13R-MnLOX, which catalyzes antarafacial hydrogen abstraction and oxygenation in analogy with sLOX-1, retained the large D-KIE and its temperature-dependent reaction rate. The kinetic differences between 13R-MnLOX and sLOX-1 may be due to protein dynamics, hydrogen donor-acceptor distances, and to the metal ligands, which may not equalize the 0.7V-gap between the redox potentials of the free metals.
材料
货号
品牌
产品描述
Sigma-Aldrich
亚油酸, ≥99%
Sigma-Aldrich
亚麻酸, ≥99%
Sigma-Aldrich
亚油酸, liquid, BioReagent, suitable for cell culture
Supelco
亚油酸, analytical standard
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
Supelco
亚麻酸, analytical standard
Sigma-Aldrich
亚油酸 钠盐, ≥98% (GC)
Sigma-Aldrich
亚油酸, technical, 58-74% (GC)
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
亚麻酸, ~70% (GC)
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
Supelco
反亚油酸 溶液, ~250 mg/mL in ethanol, analytical standard
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%