• 主页
  • 查找结果
  • A simple method to engineer a protein-derived redox cofactor for catalysis.
A simple method to engineer a protein-derived redox cofactor for catalysis.
Biochimica et biophysica acta (2014-05-27)
Sooim Shin, Moonsung Choi, Heather R Williamson, Victor L Davidson
摘要
The 6×-Histidine tag which is commonly used for purification of recombinant proteins was converted to a catalytic redox-active center by incorporation of Co(2+). Two examples of the biological activity of this engineered protein-derived cofactor are presented. After inactivation of the natural diheme cofactor of MauG, it was shown that the Co(2+)-loaded 6×His-tag could substitute for the hemes in the H2O2-driven catalysis of tryptophan tryptophylquinone biosynthesis. To further demonstrate that the Co(2+)-loaded 6×His-tag could mediate long range electron transfer, it was shown that addition of H2O2 to the Co(2+)-loaded 6×His-tagged Cu(1+) amicyanin oxidizes the copper site which is 20Å away. These results provide proof of principle for this simple method by which to introduce a catalytic redox-active site into proteins for potential applications in research and biotechnology.
材料
货号
品牌
产品描述
Sigma-Aldrich
色氨酸 -L, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
色氨酸 -L, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
色氨酸 -L, BioUltra, ≥99.5% (NT)
Supelco
色氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
色氨酸 -L, certified reference material, TraceCERT®
Sigma-Aldrich
钴, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
钴, powder, 2 μm particle size, 99.8% trace metals basis
色氨酸, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
钴, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
钴, granular, 99.99% trace metals basis
Sigma-Aldrich
钴, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
钴, pieces, 99.5% trace metals basis
Sigma-Aldrich
钴, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
钴, foil, thickness 0.25 mm, ≥99.99% trace metals basis
钴, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%
钴, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
钴, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
钴, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
钴, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
钴, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
钴, foil, 4mm disks, thickness 0.003mm, 99.9%
钴, foil, 4mm disks, thickness 0.004mm, 99.9%
钴, foil, 4mm disks, thickness 0.005mm, 99.9%
钴, foil, 4mm disks, thickness 0.006mm, 99.9%
钴, foil, 4mm disks, thickness 0.007mm, 99.9%
钴, foil, 4mm disks, thickness 0.008mm, 99.9%
钴, foil, 4mm disks, thickness 0.009mm, 99.9%
钴, foil, 4mm disks, thickness 0.0125mm, 99.9%
钴, foil, 4mm disks, thickness 0.015mm, 99.9%
钴, foil, 4mm disks, thickness 0.01mm, 99.9%