• 主页
  • 查找结果
  • Protective role of silymarin against manganese-induced nephrotoxicity and oxidative stress in rat.
Protective role of silymarin against manganese-induced nephrotoxicity and oxidative stress in rat.
Environmental toxicology (2013-01-23)
Yassine Chtourou, El mouldi Garoui, Tahia Boudawara, Najiba Zeghal
摘要
Metal toxicity may occur after exposure from many sources. Oxidative stress is thought to be involved in manganese-induced toxicity and leads to various health disorders. Silymarin (SIL), a natural flavonoid, has been reported to have many benefits and medicinal properties. The aim of this study was to assess the toxicity of manganese (Mn) on oxidative stress and DNA damage in the kidney of rats and its alleviation by SIL. Manganese was given orally in drinking water (20 mg MnCl2 /mL) with or without SIL administration (100 mg /kg intraperitoneally) for 30 days. Our data showed that SIL significantly prevented Mn induced nephrotoxicity, indicated by both diagnostic indicators of kidney injury like plasma urea, uric acid and creatinine and urinary electrolyte levels and by histopathological analysis. Moreover, Mn-induced profound elevation of the production of reactive oxygen species (ROS) and altered the levels of oxidative stress related biomarkers in kidney tissue. This is evidenced by the increase of lipid peroxidation, protein carbonylation, DNA fragmentation and urinary hydrogen peroxide, while, the activities of enzymatic antioxidant and glutathione level were decreased. Treatment with SIL reduced the alterations in the renal and urine markers, decreasing lipid peroxidation markers, increasing the antioxidant cascade and decreasing the Mn-induced damage. All these changes were supported by histopathological observations. These findings suggested that the inhibition of Mn-induced damage by SIL was due at least in part to its antioxidant activity and its capacity to modulate the oxidative damage.
材料
货号
品牌
产品描述
Sigma-Aldrich
L -还原型谷胱甘肽, ≥98.0%
Sigma-Aldrich
L -还原型谷胱甘肽, suitable for cell culture, BioReagent, ≥98.0%, powder
Supelco
谷胱甘肽, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L -还原型谷胱甘肽, BioXtra, ≥98.0%
Sigma-Aldrich
锰, powder, ≥99.9% trace metals basis
谷胱甘肽, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
锰, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
锰, chips, thickness <2.0 mm, 99%
锰, rod, 10mm, diameter 4.0mm, cast, 99.5%
锰, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
锰, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
锰, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
锰, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%