Antimicrobial activity of cobalt imidazolate metal-organic frameworks.
Chemosphere (2014-07-30)
Sonia Aguado, Jennifer Quirós, Jerome Canivet, David Farrusseng, Karina Boltes, Roberto Rosal
摘要
Two cobalt imidazolate metal-organic frameworks were evaluated as a bactericidal material against the growth of the Gram-negative bacteria Pseudomonas putida and Escherichia coli. Under the most unfavourable conditions, within the exponential growth phase and in the culture media for both microorganisms, the growth inhibition reached over 50% for concentrations of biocidal material in the 5-10mgL(-1) range. The release of metal gives excellent durability with the antibacterial effect persisting after 3months. Both cobalt-based materials can be prepared with simple, cheap and easily accessible commercial ligands, leading to a more affordable possible future application as antimicrobial materials.
材料
货号
品牌
产品描述
Sigma-Aldrich
钴, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
钴, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
钴, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
钴, granular, 99.99% trace metals basis
Sigma-Aldrich
钴, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
钴, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
钴, pieces, 99.5% trace metals basis
Sigma-Aldrich
钴, foil, thickness 0.25 mm, ≥99.99% trace metals basis
钴, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
钴, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
钴, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
钴, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
钴, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
钴, foil, 4mm disks, thickness 0.003mm, 99.9%
钴, foil, 4mm disks, thickness 0.004mm, 99.9%
钴, foil, 4mm disks, thickness 0.005mm, 99.9%
钴, foil, 4mm disks, thickness 0.006mm, 99.9%
钴, foil, 4mm disks, thickness 0.007mm, 99.9%
钴, foil, 4mm disks, thickness 0.008mm, 99.9%
钴, foil, 4mm disks, thickness 0.009mm, 99.9%
钴, foil, 4mm disks, thickness 0.0125mm, 99.9%
钴, foil, 4mm disks, thickness 0.015mm, 99.9%
钴, foil, 4mm disks, thickness 0.01mm, 99.9%
钴, foil, 4mm disks, thickness 0.025mm, 99.9%
钴, foil, 4mm disks, thickness 0.02mm, 99.9%
钴, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%
钴, foil, 4mm disks, thickness 0.05mm, as rolled, 99.9%
钴, foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%
钴, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%
钴, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%