  • Search Results
  • A central role for intermolecular dityrosine cross-linking of fibrinogen in high molecular weight advanced oxidation protein product (AOPP) formation.
A central role for intermolecular dityrosine cross-linking of fibrinogen in high molecular weight advanced oxidation protein product (AOPP) formation.
Biochimica et biophysica acta (2014-10-05)
Graziano Colombo, Marco Clerici, Daniela Giustarini, Nicola Portinaro, Salvatore Badalamenti, Ranieri Rossi, Aldo Milzani, Isabella Dalle-Donne
ABSTRACT
Advanced oxidation protein products (AOPPs) are dityrosine cross-linked and carbonyl-containing protein products formed by the reaction of plasma proteins with chlorinated oxidants, such as hypochlorous acid (HOCl). Most studies consider human serum albumin (HSA) as the main protein responsible for AOPP formation, although the molecular composition of AOPPs has not yet been elucidated. Here, we investigated the relative contribution of HSA and fibrinogen to generation of AOPPs. AOPP formation was explored by SDS-PAGE, under both reducing and non-reducing conditions, as well as by analytical gel filtration HPLC coupled to fluorescence detection to determine dityrosine and pentosidine formation. Following exposure to different concentrations of HOCl, HSA resulted to be carbonylated but did not form dityrosine cross-linked high molecular weight aggregates. Differently, incubation of fibrinogen or HSA/fibrinogen mixtures with HOCl at concentrations higher than 150 μM induced the formation of pentosidine and high molecular weight (HMW)-AOPPs (>200 k Da), resulting from intermolecular dityrosine cross-linking. Dityrosine fluorescence increased in parallel with increasing HMW-AOPP formation and increasing fibrinogen concentration in HSA/fibrinogen mixtures exposed to HOCl. This conclusion is corroborated by experiments where dityrosine fluorescence was measured in HOCl-treated human plasma samples containing physiological or supra-physiological fibrinogen concentrations or selectively depleted of fibrinogen, which highlighted that fibrinogen is responsible for the highest fluorescence from dityrosine. A central role for intermolecular dityrosine cross-linking of fibrinogen in HMW-AOPP formation is shown. These results highlight that oxidized fibrinogen, instead of HSA, is the key protein for intermolecular dityrosine formation in human plasma.
Sigma-Aldrich
L-精氨酸, reagent grade, ≥98%
Sigma-Aldrich
L-精氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Fibrinogen 来源于牛血浆, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
纤维蛋白原 来源于人类血浆, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
L -赖氨酸, ≥98% (TLC)
Sigma-Aldrich
L-酪氨酸, reagent grade, ≥98% (HPLC)
SAFC
L-精氨酸
Sigma-Aldrich
L-赖氨酸 单盐酸盐, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-精氨酸, 99%, FCC, FG
SAFC
L-酪氨酸
Sigma-Aldrich
L-赖氨酸 单盐酸盐, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-酪氨酸, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
纤维蛋白原 来源于人类血浆, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
L-酪氨酸, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L -赖氨酸, crystallized, ≥98.0% (NT)
Supelco
L-精氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-赖氨酸 单盐酸盐, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L -赖氨酸, analytical standard
精氨酸, European Pharmacopoeia (EP) Reference Standard
Supelco
L-酪氨酸, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-精氨酸, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-赖氨酸 单盐酸盐, BioUltra, ≥99.5% (AT)
酪氨酸, European Pharmacopoeia (EP) Reference Standard
USP
L-酪氨酸, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-赖氨酸 乙酸盐, ≥98% (HPLC)
Supelco
L-赖氨酸 单盐酸盐, certified reference material, TraceCERT®
赖氨酸 盐酸盐, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-酪氨酸, FG
Supelco
L-赖氨酸乙酸盐, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-酪氨酸, certified reference material, TraceCERT®