  • Search Results
  • Kinetic and thermodynamic studies on removal of Cu(II) from aqueous solutions using soil nanoclays.
Kinetic and thermodynamic studies on removal of Cu(II) from aqueous solutions using soil nanoclays.
Water environment research : a research publication of the Water Environment Federation (2015-01-30)
Yueh-Min Chen, Tsung-Ming Tsao, Ming-Kuang Wang, Shen Yu, Cheng-Chung Liu, Hong-Chun Li, Chih-Yu Chiu, Liaug-Chi Wang
ABSTRACT
Soil clays (< 2,000 nm) (SC) and soil nanoclays (< 100 nm) (SNC) were used as adsorbents for removal of Cu(II) from aqueous solution. The experiments were conducted with variables including pH, interaction time, concentration of Cu(II) and temperature. Four kinetic models have been employed to investigate adsorption mechanisms, and the experimental data more closely resemble a second-order process of the kinetic model. Adsorption studies on soil nanoclays have been shown to be highly effective in removing of Cu(II) from aqueous solution. This adsorbent is widely available as a natural material, is mechanically stable and, most importantly, it is environmentally appealing. The maximum Cu(II) adsorption capacity of soil nanoclays (31.7 mg/g) is more than three times higher than natural soil clays (10.2 mg/g). Our study demonstrates that soil nanoclays can be used effectively for removal of Cu(II) from aqueous systems to achieve environmental cleaning purposes.
Sigma-Aldrich
铜, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
铜, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
铜, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, 99.999% trace metals basis
Sigma-Aldrich
铜, nanopowder, 60-80 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
铜, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
铜, foil, ≥99.8% (complexometric)
Sigma-Aldrich
铜, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, powder, <75 μm, 99%
Sigma-Aldrich
铜, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
铜, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
铜, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
铜, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铜, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
铜, bars (random sizes), ≥99.999% trace metals basis
Sigma-Aldrich
铜, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
铜, electrolytic, Analytical Reagent, reag. Ph. Eur., ≥99.9% (complexometric), wire
Sigma-Aldrich
铜, rod, diam. 11 mm, 99.9999% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铜, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
铜, shot, −3-+14 mesh, 99%
Sigma-Aldrich
铜, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
铜, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%
Sigma-Aldrich
铜, wire, diam. 2.0 mm, 99.999% trace metals basis
铜, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
铜, mesh, 130x130mm, nominal aperture 0.2mm, thickness 0.004mm, wire diameter 0.0125mm, 118 wires/inch, open area 88%, electro-formed mesh
铜, foil, 100x100mm, thickness 0.125mm, hard, 99.95+%
铜, rod, 100mm, diameter 2.0mm, as drawn, 99.99+%